BioBasic™ 8 HPLC Columns
Actual product may vary
Thermo Scientific™

BioBasic™ 8 HPLC Columns

Use this column for reversed-phase separation of proteins and other biomolecules.
Catalog NumberLength (Metric)Diameter (Metric)
72205-01400110 mm4 mm
72205-254630250 mm4.6 mm
Catalog number 72205-014001
Price (JPY)
-
Length (Metric):
10 mm
Diameter (Metric):
4 mm

Optimized for the separation of a wide range of peptides, Thermo Scientific™ BioBasic™ 8 LC Columns are high-performance columns ideal for the analysis of proteins, peptides and other biomolecules.

  • 300Å pore size for improved biomolecule analysis
  • An excellent starting column for protein and peptide analysis
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations

Ordering Information:

Other column dimensions are available, including preparative columns. Please call Customer Service for more information.

Specifications
Column FormatGuard Cartridge, 4 Pk.
Column TypeReversed Phase
EndcappedYes
For Use WithUniguard Direct-Connection and Stand Alone Holder
Max. Pressure5800 psi (400 bar)
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Stationary PhaseC8
Surface Area100 m²/g
Temperature60°C
USP TypeL7
Carbon Load5%
Diameter (Metric)4 mm
Length (Metric)10 mm
Particle Size5 μm
PhaseReversed Phase
Pore Size300 Å
Product LineBioBasic
TypeGuard Cartridge
pH2 to 9
Unit SizePack of 4