BioBasic™ 8 HPLC Columns
Thermo Scientific™

BioBasic™ 8 HPLC Columns

Use this column for reversed-phase separation of proteins and other biomolecules.
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric)
72205-254630250 mm4.6 mm
Catalog number 72205-254630
Price (EUR)
960,43
Each
Length (Metric):
250 mm
Diameter (Metric):
4.6 mm
Price (EUR)
960,43
Each

Optimized for the separation of a wide range of peptides, Thermo Scientific™ BioBasic™ 8 LC Columns are high-performance columns ideal for the analysis of proteins, peptides and other biomolecules.

  • 300Å pore size for improved biomolecule analysis
  • An excellent starting column for protein and peptide analysis
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations

Ordering Information:

Other column dimensions are available, including preparative columns. Please call Customer Service for more information.

Specifications
Column FormatAnalytical Column
Column TypeReversed Phase
EndcappedYes
Max. Pressure5800 psi (400 bar)
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Product TypeAnalytical Column
Stationary PhaseC8
Surface Area100 m²/g
Temperature60°C
USP TypeL7
Carbon Load5%
Diameter (Metric)4.6 mm
Length (Metric)250 mm
Particle Size5 μm
PhaseReversed Phase
Pore Size300 Å
Product LineBioBasic
TechniqueBio-LC
pH2 to 9
Unit SizeEach