Nalgene™ Syringe Filters
Nalgene™ Syringe Filters
Nalgene™ Syringe Filters
Nalgene™ Syringe Filters
Thermo Scientific™

Nalgene™ Syringe Filters

Filter sample volumes of 10 to 100mL with these sterile syringe filters featuring a variety of membranes and pore sizes for most laboratory applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPore SizeDiameter (Metric)Material (Membrane)Sterility
723-99200.2 μm25 mmSFCASterile
720-13200.2 μm13 mmPESSterile
722-25200.2 μm25 mmGlass Fiber, Cellulose AcetateSterile
723-25200.2 μm25 mmSFCASterile
723-25450.45 μm25 mmSFCASterile
723-25800.8 μm25 mmCellulose AcetateSterile
723-99450.45 μm25 mmSFCASterile
725-25200.2 μm25 mmPESSterile
725-25450.45 μm25 mmPESSterile
726-25200.2 μm25 mmNylonSterile
726-25450.45 μm25 mmNylonSterile
171-00200.2 μm4 mmCellulose AcetateNon-sterile
171-00450.45 μm4 mmCellulose AcetateNon-sterile
Catalog number 723-9920
Price (MXN)
-
Pore Size:
0.2 μm
Diameter (Metric):
25 mm
Material (Membrane):
SFCA
Sterility:
Sterile

Thermo Scientific™ Nalgene™ syringe filters are built to meet your lab’s clarification and sterilization needs. Our filters are available in a variety of sizes and membranes that can be easily used for both sterile and non-sterile laboratory applications. Whether your research necessitates prefiltration, sample preparation, sterile filtration, laboratory filtration, or gas venting, Nalgene syringe filters are lab essentials.

Key Features
  • 10 to 100mL sample size
  • Inlet: Female Luer-Lok™
  • Outlet: Male Luer slip
  • Polypropylene housing with ID color rings
  • Pore size and membrane type printed on unit
Polyether Sulfone (PES)
  • Highest throughput and lowest clogging
  • Low binding
  • Most commonly used general purpose membrane
Surfactant-free Cellulose Acetate (SFCA)
  • Low protein binding, lower extractables than standard CA
  • Good for cell culture fluids and cellular solutions
  • Polypropylene housings with blue color rings
Cellulose Acetate (CA)
  • Some protein binding and extractables
  • For sterile filtration and clarification of: buffers, aqueous solutions and reagents
  • Polypropylene housings with red color rings
Nylon
  • Naturally hydrophilic, low extractables, some protein binding
  • Useful for alcohols, cryoprotectants such as DMSO and with aqueous/biological and weak solvents
  • Polypropylene housings with pink color rings
PTFE
  • Naturally hydrophobic
  • Excellent for HPLC/GC solvent filtration. Ideal for sterile venting of air or other gases during filtration
  • Polypropylene housings with white color rings

Specifications
Diameter (Metric)25 mm
For Use With (Application)Sterilization and clarification of biological/immunological samples, cell culture fluids, cellular solutions, and biological fluids
Green FeaturesFewer resources used and less waste
Housing MaterialPolypropylene
Quantity125/Pk.
TypeSyringe Filter
Volume (Metric)10 to 100 mL (Sample)
Filter TypeMembrane Filter
Material (Membrane)SFCA
Pore Size0.2 μm
SterilitySterile
Unit SizeCase of 125

Frequently asked questions (FAQs)

Are Nalgene Sterile Syringe Filters packed individually or in bulk?

The Nalgene Sterile Syringe Filters are individually blister-packed. The Nalgene non-Sterile Syringe Filters are bulk packed.