BioBasic™ 4 HPLC Columns
Actual product may vary

BioBasic™ 4 HPLC Columns

Use this 300Å wide pore silica for analysis of larger hydrophobic proteins and peptides that bind too tightly on C8 or C18 columns.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)
72305-05213050 mm2.1 mm
72305-05103050 mm1 mm
72305-101030100 mm1 mm
72305-102130100 mm2.1 mm
72305-104630100 mm4.6 mm
72305-152130150 mm2.1 mm
72305-154630150 mm4.6 mm
72305-252130250 mm2.1 mm
72305-254630250 mm4.6 mm
72305-01210110 mm2.1 mm
72305-01400110 mm4 mm
Catalog number 72305-052130
HSN 90279090
Price (INR)
77,266.00
Each
Length (Metric):
50 mm
Diameter (Metric):
2.1 mm
Price (INR)
77,266.00
Each

Designed for the analysis of larger proteins and peptides, Thermo Scientific™ BioBasic™ 4 LC Columns are high-performance columns ideal for the analysis of proteins, peptides and other biomolecules with high hydrophobicity.

  • Based on 300Å silica for outstanding biomolecule performance
  • Lower carbon load for optimal retention of larger peptides and proteins
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations

Ordering Information:

Other column dimensions are available, including preparative columns. Please call your Customer Service Representative for more information.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Length (Metric)50 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 9
Pore Size300 Å
Surface Area100 m²/g
Temperature60°C
USP TypeL26
Stationary PhaseC4
Product LineBioBasic
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load4%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
TypeAnalytical Column
Unit SizeEach