BioBasic™ 4 HPLC Columns
Thermo Scientific™

BioBasic™ 4 HPLC Columns

Use this 300Å wide pore silica for analysis of larger hydrophobic proteins and peptides that bind too tightly on C8 or C18 columns.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호길이(미터법)직경(미터법)
72305-102130100 mm2.1 mm
72305-05103050 mm1 mm
72305-05213050 mm2.1 mm
72305-101030100 mm1 mm
72305-104630100 mm4.6 mm
72305-152130150 mm2.1 mm
72305-154630150 mm4.6 mm
72305-252130250 mm2.1 mm
72305-254630250 mm4.6 mm
72305-01210110 mm2.1 mm
72305-01400110 mm4 mm
카탈로그 번호 72305-102130
제품 가격(KRW)
-
길이(미터법):
100 mm
직경(미터법):
2.1 mm

Designed for the analysis of larger proteins and peptides, Thermo Scientific™ BioBasic™ 4 LC Columns are high-performance columns ideal for the analysis of proteins, peptides and other biomolecules with high hydrophobicity.

  • Based on 300Å silica for outstanding biomolecule performance
  • Lower carbon load for optimal retention of larger peptides and proteins
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations

Ordering Information:

Other column dimensions are available, including preparative columns. Please call your Customer Service Representative for more information.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
사양
컬럼 형식Analytical Column
컬럼 유형Reversed Phase
엔드캡 처리됨Yes
최대 압력5800 psi (400 bar)
팩킹 물질Spherical, Fully Porous, Ultrapure Silica
정지상C4
표면적100 m²/g
온도60°C
USP 유형L26
탄소 부하4%
직경(미터법)2.1 mm
길이(미터법)100 mm
입자 크기5 μm
Reversed Phase
기공 크기300 Å
제품라인BioBasic
유형Analytical Column
pH2 to 9
Unit SizeEach