Manostat™ Norprene™ Pumping Links
Actual product may vary
Thermo Scientific™
Manostat™ Norprene™ Pumping Links
Manostat norprene pumping links are for use with Manostat pumps and dispensers.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호튜빙 크기설명
7230513513Simon/Vera, Kate and Compulab RD2
7530525525Preston, Spencer and Compulab RD1
7530514514Preston, Spencer and Compulab RD1
7530516516Preston, Spencer and Compulab RD1
7530513513Preston, Spencer and Compulab RD1
7530517517Preston, Spencer and Compulab RD1
7230514514Simon/Vera, Kate and Compulab RD2
7230525525Simon/Vera, Kate and Compulab RD2
7230516516Simon/Vera, Kate and Compulab RD2
7230517517Simon/Vera, Kate and Compulab RD2
카탈로그 번호 72305135
제품 가격(KRW)
-
튜빙 크기:
13
설명:
Simon/Vera, Kate and Compulab RD2
Thermo Scientific™ Manostat norprene pumping links are for use with Manostat pumps and dispensers.
사양
설명Simon/Vera, Kate and Compulab RD2
용도Simon/Vera, Kate and Compulab RD2
포함5 links/pack
제품라인Thermo Scientific
튜빙 크기13
직경(영국식 단위) 내부1/32 in.
직경(영국식 단위) 외부0.507 in.
직경(미터법) 내부0.8 mm
직경(미터법) 외부13 mm
물질Norprene
제품 유형Pumping Link
Unit SizeEach