Thermo Scientific™
Richard-Allan Scientific™ Signature Series Hematoxylin 2 7231
Related applications:
Clinical
Yield visually similar results to Gills 2 and 3 with Thermo Scientific™ Richard-Allan Scientific™ Signature Series Hematoxylin 2, a uniquely formulated product.
Back to top
  • No filtration required
  • Stains up to 2500 slides per pint
  • Part of the Signature Series Staining System
  • Crisp, well-defined nuclear detail chromatin delineation
Available in 5L packaging
  • Reduces plastic by 90%
  • Increases stain stability
  • Reduces messy stain spills
  • Saves 35% storage space versus traditional packaging