Nalgene™ Syringe Filters, Non-sterile
Thermo Scientific™

Nalgene™ Syringe Filters, Non-sterile

Filter sample volumes of 10 to 100mL with these nonsterile syringe filters featuring a variety of membranes and pore sizes for most laboratory applications.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호기공 크기직경(미터법)재질(멤브레인)제품 사이즈
724-2045PK
724-2045으로도 사용됨
0.45 μm25 mmSFCAPack of 100
721-13200.2 μm13 mmPTFECase of 100
721-13450.45 μm13 mmPTFECase of 100
722-20001.1 μm25 mmGlass FiberCase of 50
724-20200.2 μm25 mmSFCACase of 500
724-2020PK
724-2020으로도 사용됨
0.2 μm25 mmSFCAPack of 100
724-20450.45 μm25 mmSFCACase of 500
724-20800.8 μm25 mmCellulose AcetateCase of 500
724-2080PK
724-2080으로도 사용됨
0.8 μm25 mmCellulose AcetatePack of 100
727-20200.2 μm25 mmNylonCase of 500
727-2020PK
727-2020으로도 사용됨
0.2 μm25 mmNylonPack of 100
727-20450.45 μm25 mmNylonCase of 500
727-2045PK
727-2045으로도 사용됨
0.45 μm25 mmNylonPack of 100
728-20200.2 μm25 mmPTFECase of 500
728-2020PK
728-2020으로도 사용됨
0.2 μm25 mmPTFEPack of 100
728-20450.45 μm25 mmPTFECase of 500
728-2045PK
728-2045으로도 사용됨
0.45 μm25 mmPTFEPack of 100
176-00200.2 μm4 mmNylonCase of 400
176-0020PK
176-0020으로도 사용됨
0.2 μm4 mmNylonPack of 100
176-00450.45 μm4 mmNylonCase of 400
176-0045PK
176-0045으로도 사용됨
0.45 μm4 mmNylonPack of 100
카탈로그 번호 724-2045PK
724-2045으로도 사용됨
제품 가격(KRW)
442,000
Pack of 100
기공 크기:
0.45 μm
직경(미터법):
25 mm
재질(멤브레인):
SFCA
Unit Size:
Pack of 100
제품 가격(KRW)
442,000
Pack of 100

Thermo Scientific™ Nalgene™ syringe filters are built to meet your lab’s clarification and sterilization needs. Our filters are available in a variety of sizes and membranes that can be easily used for both sterile and non-sterile laboratory applications. Whether your research necessitates prefiltration, sample preparation, sterile filtration, laboratory filtration, or gas venting, Nalgene syringe filters are lab essentials.

Key Features
  • 10 to 100mL sample size
  • Inlet: Female Luer-Lok™
  • Outlet: Male Luer slip
  • Polypropylene housing with ID color rings
  • Pore size and membrane type printed on unit

Surfactant-free Cellulose Acetate (SFCA)

  • Low protein binding, lower extractables than standard CA
  • Good for cell culture fluids and cellular solutions
  • Polypropylene housings with blue color rings

Cellulose Acetate (CA)

  • Some protein binding and extractables
  • For sterile filtration and clarification of: buffers, aqueous solutions and reagents
  • Polypropylene housings with red color rings

Nylon

  • Naturally hydrophilic, low extractables, some protein binding
  • Useful for alcohols, cryoprotectants such as DMSO and with aqueous/biological and weak solvents
  • Polypropylene housings with pink color rings
PTFE
  • Naturally hydrophobic
  • Excellent for HPLC/GC solvent filtration. Ideal for sterile venting of air or other gases during filtration
  • Polypropylene housings with white color rings

사양
직경(미터법)25 mm
직경(미터법) 멤브레인25 mm
용도(애플리케이션)Filtration of biological/immunological samples, cell culture fluids, cellular solutions, and biological fluids
그린 기능Fewer resources used and less waste
하우징 재질Polypropylene (Housing)
제품라인Nalgene
유형Syringe Filter
부피(미터법)10 to 100 mL (Sample)
재질(멤브레인)SFCA
기공 크기0.45 μm
멸균Non-sterile
Unit SizePack of 100