Thermo Scientific™
Richard-Allan Scientific™ Gill™ 1, 2, 3 Hematoxylin
Related applications:
Clinical
With three strengths available, the Thermo Scientific™ Richard-Allan Scientific™ Gill™ 1, 2, 3 Hematoxylin allows complete control of staining intensities.
Back to top
Gill Hematoxylins are progressive nuclear stains.
  • Gill 1 is recommended for use in cytology
  • Gill 2 has applications in histology and cytology
  • Gill 3 is used in histology
  • All are pre-mixed and ready-to-use