MEN1 Antibody (4221)

Catalog number: 730012

Invitrogen™  Related applications: Protein Assays and Analysis | Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

MEN1 Monoclonal Antibody for IP
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reactivity: Human
Host Species: Mouse
SubClass (Isotype): IgG
Clonality: Monoclonal
Clone ID: 4221
Antibody Fragment: Whole Antibody
Target Gene: MEN1
Gene ID: 4221
Gene Symbol: MEN1
Accession Number: O00255
Label or Dye: Unconjugated
Purification: Purified
Concentration: 0.5 mg⁄ml
Form: Liquid
Mass: 100 µg
Neutralizing Antibody: No
Product Size: 100 µg
Volume (Metric): 200 µL
Shipping Condition: Wet Ice

Contents & storage

2 - 8°C for up to 1 month, -20°C for long storage.



Avoid repeated freezing and thawing.

Documents

Manuals & protocols

Recommended products