SMYD3 Antibody

Catalog number: 730061

Invitrogen™  Related applications: Protein Assays and Analysis | Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

SMYD3 is a histone methyltransferase that plays a role in transcriptional regulation as a member of an RNA polymerase complex.

Specifications

Product Size: Pack of 5
For Use With: Splitless Liner with Recessed Gooseneck
Item Description: 453A2325
Packing Material: None
Diameter (Metric) Inner: 2mm
Diameter (Metric) Outer: 6.3mm
Length (Metric): 78.5mm
For Use With (Equipment): GC Injection Port
Quantity: 5

Contents & storage

2 - 8 °C for up to 1 month, -20 °C for long storage.

Avoid repeated freezing and thawing.

Documents

Manuals & protocols

Recommended products