CaptureSelect™ HRP Anti-C-tag Conjugate
Thermo Scientific™

CaptureSelect™ HRP Anti-C-tag Conjugate

Thermo Scientific™ CaptureSelect™ HRP Anti-C-tag Conjugate consists of a 13-kDa recombinant single-domain antibody fragment (VHH affinity ligand) with high affinity and selectivity for the four-amino-acid 'C-tag' peptide tag E‐P‐E‐A (glutamic acid-proline-glutamic acid-alanine).
Have Questions?
Número de catálogoCantidad
7303072100100 μL
Número de catálogo 7303072100
Precio (USD)
-
Cantidad:
100 μL

CaptureSelect HRP Anti-C-tag Conjugate consists of a 13-kDa recombinant single-domain antibody fragment (VHH affinity ligand) with high affinity and selectivity for the four-amino-acid 'C-tag' peptide tag E‐P‐E‐A (glutamic acid-proline-glutamic acid-alanine). The anti-C-tag affinity ligand recognizes the E-P-E-A peptide when this tag is fused directly to the C-terminus of a protein. The affinity ligand is chemically conjugated to horseradish peroxidase (HRP) via an appropriate spacer that retains the binding reactivity of the ligand.

Applications for CaptureSelect HRP Anti-C-tag Conjugate include detection of C-tag fusion proteins in direct ELISA or western blot applications.

  • Format type: HRP, liquid
  • Buffer: PBS, 50% (v/v) glycerol
  • Storage: in freezer (-5 to -30°C)
  • Concentration: 0.5 mg/mL
  • Volume: 0.1 mL
  • Recommended working dilution for ELISA and western blot: 1:1000 to 1:5000 (the optimal working dilution should be determined empirically for each application)

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12- to 15-kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Especificaciones
Para utilizar con (aplicación)Quantitation of C-tag tagged proteins
Cantidad100 μL
ObjetivoE-P-E-A (glutamic acid - proline - glutamic acid - alanine)
Capacidad (métrico)100 μL
FormularioLíquido
Línea de productosCaptureSelect
TipoConjugate
Unit SizeEach