CaptureSelect™ HRP Anti-CD81 Conjugate
Thermo Scientific™

CaptureSelect™ HRP Anti-CD81 Conjugate

Thermo Scientific™ CaptureSelect™ HRP Anti-CD81 Conjugate includes a 12–15-kDa recombinant single-domain antibody fragment (VHH affinity ligand) that specifically binds with high affinity to the tetraspanin marker CD81.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
73039821000.1 mL
73039825000.5 mL
Catalog number 7303982100
Price (MXN)
-
Quantity:
0.1 mL

CaptureSelect HRP Anti-CD81 Conjugate includes a 12–15-kDa recombinant single-domain antibody fragment (VHH affinity ligand) that specifically binds with high affinity to the tetraspanin marker CD81. The affinity ligand is chemically conjugated to horseradish peroxidase (HRP) via an appropriate spacer to maintain full binding capacity. This conjugate allows you to detect, quantitate, and characterize CD81-expressing extracellular vesicles (EVs) like exosomes.

CaptureSelect HRP Anti-CD81 Conjugate can be combined with CaptureSelect Biotin Anti-CD81 Conjugate (Cat. No. 7103982100) in enzyme-linked immunoassays to quantify the CD81-expressing EVs. Sandwich ELISA setups can be designed using the biotin anti-CD81 conjugate as a capture reagent immobilized on a streptavidin-coated surface. HRP anti-CD81 conjugate is then used for detection of bound CD81-positive EVs.

For detection of a broad variety of EVs and exosomes that originate from different cell types and secretion pathways, such as those of endosomal (CD63-enriched, CD9+, or CD81+) or ectosomal (CD63 low, CD9+, and CD81+) origin, we also offer CaptureSelect Biotin and HRP anti-CD63 conjugates and CaptureSelect Biotin and HRP anti-CD9 conjugates.

Recommended working dilution for sandwich ELISA: 1:5000 to 1:10,000 (the optimal working dilution should be determined empirically for each application).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration0.5 mg/mL
Final Product TypeHRP Conjugate
FormulationPBS, 50% (v/v) glycerol
Label or DyeHorseradisch peroxidase (HRP)
Ligand TypeRecombinant single-domain antibody fragment (VHH ligand)
Quantity0.1 mL
ReactivityAnti-tetraspanin CD81
Shipping ConditionDry Ice
TargetTetraspanin CD81
FormLiquid
Product LineCaptureSelect
Unit SizeEach
Contents & Storage
Store in freezer (–5 to –30°C).