BioBasic™ AX HPLC Columns
Thermo Scientific™

BioBasic™ AX HPLC Columns

Use Thermo Scientific™ BioBasic™ AX LC Columns for the separation of proteins, peptides, other anionic species and highly polar molecules.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)
73105-05103050 mm1 mm
73105-05213050 mm2.1 mm
73105-05303050 mm3 mm
73105-05463050 mm4.6 mm
73105-101030100 mm1 mm
73105-102130100 mm2.1 mm
73105-104630100 mm4.6 mm
73105-151030150 mm1 mm
73105-152130150 mm2.1 mm
73105-153030150 mm3 mm
73105-154630150 mm4.6 mm
73105-254630250 mm4.6 mm
73105-01210110 mm2.1 mm
Catalog number 73105-051030
Price (BRL)
-
Length (Metric):
50 mm
Diameter (Metric):
1 mm

Use Thermo Scientific™ BioBasic™ AX LC Columns for the separation of proteins, peptides, other anionic species and highly polar molecules.

  • 5μm particle size, 300Å pore size
  • Proprietary, protein-friendly, ion exchange ligands of polyethyleneimine (PEI) bound to pure deactivated silica
  • Low hydrophobicity
  • Capacity of 0.22mEq/g; 3% carbon load
  • Use for normal phase and hydrophilic interaction chromatography (HILIC)
  • Stable under demanding pH conditions (2 to 8 pH range)
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatAnalytical Column
Column TypeMixed Mode
EndcappedNo
HydrophobicityLow
Max. Pressure5800 psi (400 bar)
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Stationary PhasePolyethyleneimine
Surface Area100 m²/g
Temperature60°C
USP TypeN/A
Carbon Load3%
Diameter (Metric)1 mm
Length (Metric)50 mm
Particle Size5 μm
PhaseNormal Phase, HILIC, Ion Exchange
Pore Size300 Å
Product LineBioBasic
TechniqueBio-LC
pH2 to 8
Unit SizeEach