Search
Search
| Catalog Number | Length (Metric) | Diameter (Metric) |
|---|---|---|
| 73205-101030 | 100 mm | 1 mm |
| 73205-052130 | 50 mm | 2.1 mm |
| 73205-102130 | 100 mm | 2.1 mm |
| 73205-104630 | 100 mm | 4.6 mm |
| 73205-151030 | 150 mm | 1 mm |
| 73205-152130 | 150 mm | 2.1 mm |
| 73205-251030 | 250 mm | 1 mm |
| 73205-014001 | 10 mm | 4 mm |
Use Thermo Scientific™ BioBasic™ SCX LC Columns for the optimized separation of proteins, peptides and other cationic species. These high-performance columns are ideal for the analysis of proteins, peptides and other biomolecules with higher efficiency than polymeric ion exchange.
Ordering Information:
Other column dimensions are available. Please call Customer Service for more information.