BioBasic™ PREP SEC 60 Size Exclusion Chromatography HPLC Columns
Actual product may vary
Thermo Scientific™

BioBasic™ PREP SEC 60 Size Exclusion Chromatography HPLC Columns

Achieve outstanding size separation of water soluble compounds with BioBasic™ PREP SEC 60 Size Exclusion Chromatography HPLC Columns.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Column Format
73305-037821A30 mmPreparative Guard Column
73305-157846A150 mmPreparative Column, Standard
73305-307846A300 mmPreparative Column, Standard
Catalog number 73305-037821A
Price (TWD)
-
Length (Metric):
30 mm
Column Format:
Preparative Guard Column

Provide outstanding size separation of water soluble compounds with Thermo Scientific™ BioBasic™ SEC 60 Size Exclusion Chromatography preparative HPLC Columns. BioBasic SEC columns provide high efficiency separations for a wide range of molecular weights from 100 to 60,000 daltons, allowing for the analysis of proteins, peptides and other biomolecules.

BioBasic™ PREP SEC 60 HPLC Column Benefits:

  • Covers separation of analytes over a wide molecular weight range
  • Long column life and high column efficiencies
  • Simple mechanism of interaction based on molecular size and shape
  • Excellent for sample cleanup
  • Straightforward method development, simple mobile phases
  • 60Å pore size

Ordering Information:

For information on bulk quantities, please inquire.

Specifications
Column FormatPreparative Guard Column
Column TypeSize Exclusion
EndcappedNo
For Use WithPreparative Size Exclusion
For Use With (Application)Preparative Chromatography
Max. Pressure4300 psi (295 bar)
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Stationary PhaseDiol (OH)
Surface Coverage450 m²/g
Temperature60°C
USP TypeL59
Carbon Load>9%
Diameter (Metric)7.8 mm
Length (Metric)30 mm
Particle Size5 μm
PhaseSize Exclusion
Pore Size60 Å
pH2 to 8
Unit SizeEach