BioBasic™ PREP SEC 1000 Size Exclusion Chromatography HPLC Columns
Actual product may vary
Thermo Scientific™

BioBasic™ PREP SEC 1000 Size Exclusion Chromatography HPLC Columns

Achieve outstanding size separation of water soluble compounds with BioBasic™ PREP SEC 1000 Size Exclusion Chromatography HPLC Columns.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)
73605-307846A300 mm
73605-037821A30 mm
73605-157846A150 mm
Catalog number 73605-307846A
Price (TWD)
-
Length (Metric):
300 mm

Provide outstanding size separation of water soluble compounds with Thermo Scientific™ BioBasic™ SEC 1000 Size Exclusion Chromatography preparative HPLC Columns. BioBasic SEC columns provide high efficiency separations for a wide range of molecular weights from 70,000 to 10,000,000 daltons, allowing for the analysis of proteins, peptides and other biomolecules.

BioBasic™ PREP SEC 1000 HPLC Column Benefits:

  • Covers separation of analytes over a wide molecular weight range
  • Long column life and high column efficiencies
  • Simple mechanism of interaction based on molecular size and shape
  • Excellent for sample cleanup
  • Straightforward method development, simple mobile phases
  • 1000Å pore size

Ordering Information:

For information on bulk quantities, please inquire.

Specifications
Column FormatPreparative Column, Standard
Column TypeSize Exclusion
EndcappedNo
For Use WithPreparative Size Exclusion
Max. Pressure4300 psi (295 bar)
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Stationary PhaseDiol (OH)
Surface Area25 m²/g
Temperature60°C
USP TypeL59
Carbon Load>0.6%
Diameter (Metric)7.8 mm
Length (Metric)300 mm
Particle Size5 μm
PhaseSize Exclusion
Pore Size1000 Å
pH2 to 8
Unit SizeEach