Rheodyne™ Replacement Rotor Seals for Injectors
Actual product may vary
Thermo Scientific™
Rheodyne™ Replacement Rotor Seals for Injectors
Maintain performance of your Thermo Scientific™ Rheodyne™ Sample Injectors for HPLC with these replacement rotor seals.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Material
7410-075Rheodyne Injector 7410ETFE
3725-018Rheodyne Injectors 3725/3725i/3725-038/3725i-038PEEK
7010-039Rheodyne Injectors 7000/7010/7040/7067Vespel
7010-071Rheodyne Injectors 7000/7010/7040ETFE
7030-003Rheodyne Injector 7030Vespel
7125-047Rheodyne Injectors 7125/7126Vespel
Catalog number 7410-075
Price (ZAR)
-
For Use With (Equipment):
Rheodyne Injector 7410
Material:
ETFE
Use Thermo Scientific™ Rheodyne™ Replacement Rotor Seals for Injectors to maintain the performance of your Rheodyne Sample Injectors for high-performance liquid chromatography (HPLC). These seals are suitable for popular Rheodyne injector models and are available in Vespel, PEEK or ETFE.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description7410-075
MaterialETFE
For Use With (Equipment)Rheodyne Injector 7410
TypeReplacement Rotor Seals
Unit SizeEach