PCR Markers, 50-2,000 bp
Catalog number: 74601250
Invitrogen™  Related applications: Diagnostic Testing
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
PCR Markers are double-stranded linear DNA bands in gel loading buffer. The middle bands have a stronger intensity for easy orientation. Ready to use. For use in agarose gel electrophoresis for accurate analysis of PCR product sizes.
Contents & storage
Store in refrigerator (2–8°C).
Documents
Manuals & protocols
Recommended products