H-Flex 1  Tissue Culture Package
H-Flex 1  Tissue Culture Package
H-Flex 1  Tissue Culture Package
H-Flex 1  Tissue Culture Package
H-Flex 1  Tissue Culture Package
H-Flex 1 Tissue Culture Package
H-Flex 1 rotor cell culture package offers the flexibility to tun microplates, 50 ml and 15 ml conicals in the same rotor.
Have Questions?
Catalog number 75000134
Price (MXN)
-
H-Flex 1 rotor blood processing package containing adapters for microplates, 50 mL and 15 mL conicals
H-Flex 1 rotor tissue culture package containing adapters for microplates, 50 mL and 15 mL conicals.
Specifications
Certifications/ComplianceBuckets are available with biocontainment certified lids
DescriptionH-Flex 1 rotor  Cell Culture Package
Includes
  • 75003300 H-Flex 1 Rotor
  • 75003301 H-Flex 1 Buckets
  • 75003302 H-Flex 1 Lids
  • 75003307 Set of 2 Microplate Carriers, 5 standard, 1 deepwell
  • 75003303 Set of 2 adapters 8 x 50 ml conical
  • 75003304 Set of 2 adapters 18 x 15ml conical 
Product TypeCan be used with Thermo Scientific Sorvall Legend X1, X1 Pro and ST16, ST1 Plus Series, Thermo Scientific Multifuge X1, X1 Pro and Megafuge 16, ST1 Plus Series, Thermo Scientific SL 16, ST1 Plus Series
Capacity2x10 standard microplates (2x1 deepwell), 16 x 50 mL, 36 x 15 mL conicals
Max. RCF4,700 x g
Max. Speed4297
Product LineSorvall
Unit SizeEach