DNA Grade Agarose
Catalog number: 75000500
Invitrogen™  Related applications: Diagnostic Testing
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
DNA Grade Agarose has a high sieve capacity, providing optimal resolution of DNA during gel electrophoresis. The low sulfur content allows gels to be run faster without becoming brittle. Supplied as a powder that is reconstituted in 0.5X TBE for high-quality agarose gels.
Contents & storage
Store at room temperature.
Documents
Manuals & protocols
Recommended products