Denhardt's Solution (50X)
Invitrogen™

Denhardt's Solution (50X)

Denhardt's Solution is a mixture of blocking agents used in membrane-based hybridization protocols. The solution contains 1% Ficoll (type 400),자세히 알아보기
Have Questions?
카탈로그 번호수량
750018100 mL
카탈로그 번호 750018
제품 가격(KRW)
264,000
온라인 행사
Ends: 30-Sep-2026
301,000
할인액 37,000 (12%)
Each
수량:
100 mL
대량 주문 또는 맞춤형 요청
제품 가격(KRW)
264,000
온라인 행사
Ends: 30-Sep-2026
301,000
할인액 37,000 (12%)
Each
Denhardt's Solution is a mixture of blocking agents used in membrane-based hybridization protocols. The solution contains 1% Ficoll (type 400), 1% polyvinylpyrrolidone, and 1% bovine serum albumin. Sterile filtered.
For Research Use Only. Not for use in diagnostic procedures.
사양
버퍼Blotting Buffer
제품라인UltraPure
제품 유형Denhardt's Solution
수량100 mL
배송 조건Room Temperature
Unit SizeEach
구성 및 보관
Store in freezer (-5 to -30°C).