Pico™ 17 Microcentrifuge
Imagem ilustrativa. O produto pode variar
Thermo Scientific™
Pico™ 17 Microcentrifuge
The compact footprint of this microcentrifuge is ideal for routine laboratory work up to 17,000 xg.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoElectrical RequirementsIncludesDescription
75002411120 V 60 Hz24 x 1.5/2.0mL Rotor ClickSeal Biocontainment LidPico 17 Centrifuge, Ventilated
75002401120 V 60 HzDual Row 18 x 2.0/0.5mL rotor with screw-on lidPico 17 Centrifuge, Ventilated
75002414230 V 50/60 HzDual Row 18 x 2.0/0.5mL rotor with screw-on lidPico 17 Centrifuge, Ventilated
75002410230 V 50/60 Hz24 x 1.5/2.0mL Rotor ClickSeal Biocontainment LidPico 17 Centrifuge, Ventilated
75002492120 V 60 HzHematocrit rotorPico 17 Centrifuge, Ventilated
Número do catálogo 75002411
Preço (BRL)
-
Electrical Requirements:
120 V 60 Hz
Includes:
24 x 1.5/2.0mL Rotor ClickSeal Biocontainment Lid
Description:
Pico 17 Centrifuge, Ventilated
Ideal for everyday laboratory use, Thermo Scientific Pico 17 microcentrifuges provide outstanding processing power, unique safety features and a broad array of microcentrifuge rotors and accessories for microcentrifuge tube processing.
Support your micro-volume separation needs with:
  • Performance up to 17,000 x g with fast acceleration and deceleration.
  • Choose from 7 optional microcentrifuge rotors to find the ideal fit for your application:
    • Outstanding corrosion resistance with lightweight engineered polymer rotors.
    • Standard rotor runs 24 microcentrifuge tubes in a single row, from 1.5 to 2.0 mL tubes to mini-preps and spin columns.
    • Ergonomic one-click rotor opening and closing, as well as transparent design for added safety and convenience with Thermo Scientific ClickSeal biocontainment lids.
    • Eliminate the need for adapters with unique dual row rotor.
  • Simplify operation with intuitive controls, easy-to-read displays and fast one-click centrifuge lid closure.
  • Sleek, lightweight, space-saving design.
  • Durable and easy to clean and maintain.
  • Quiet performance promotes a safe, stress-free environment.
  • Conforms to the latest clinical and safety standards, including 98/79/EC In Vitro Diagnostic Directive.
Explore our clinical centrifuge portfolio for diverse clinical applications.
Learn more
Especificações
Certifications/ComplianceCSA, CE, IVD
DescriptionPico 17 Centrifuge, Ventilated
Dimensions (HWD)8.9 x 9.6 x 13.8 in. (22.5 x 24.3 x 35.2 cm)
Electrical Requirements120 V 60 Hz
Frequency60 Hz
Includes24 x 1.5/2.0mL Rotor ClickSeal Biocontainment Lid
Net Weight23.1lb. (10.5 kg)
Noise Level56 dB(A)
Product TypeBenchtop Centrifuge
Rotor24 x 1.5/2 mL Rotor with Clickseal Biocontainment Lid
Run Time1 to 99 min. (1 min. increments); HOLD
StandardsIEC 61010-1, IEC 61010-2-020 (refer to manual for more details)
Temperature Range (Metric)Ambient temperature
Voltage120 V
Capacity24 x 1.5/2 mL
Max. RCF17,000 x g
Max. Speed13,300 rpm
Product LinePico
RefrigeratedNo
Unit SizeEach