TCF-20 Zonal Rotor
Thermo Scientific™
TCF-20 Zonal Rotor
Achieve high g-force large volume gradient separations with the Thermo Scientific™ TCF-20 Zonal Rotor.
Catalog NumberCapacity
750030131350 mL
Catalog number 75003013
Price (GBP)
-
Capacity:
1350 mL
Achieve high g-force large volume gradient separations with the Thermo Scientific™ TCF-20 Zonal Rotor. Rotor innovations—from Thermo Scientific™ Auto-Lock™ rotor exchange to Thermo Scientific™ Auto-ID™ instant rotor identification and speed handles on each rotor lid—shorten run set-up time while providing peace-of-mind that the rotor is secure.
Auto-Lock Rotor Exchange
  • Secure, push-button rotor exchange in less than 3 seconds
  • Improved safety and convenience
  • No tools required
  • Quickly change rotors and applications
Auto-ID Instant Rotor Identification
  • Immediate identification of a rotor
  • Rotor specifications automatically loaded into centrifuge parameters
  • Shortens run set-up time, improves safety
Speed Handle
  • Accelerates and simplifies rotor lid tightening, ensuring lid is properly attached
  • Easier and safer lifting and carrying of rotors, further enhanced with the lightweight design
Specifications
DescriptionTCF-20 Zonal Rotor
For Use With (Equipment)Sorvall LYNX 6000 Superspeed Centrifuge
K Factor826
Net Weight17.2 lb. (7.8 kg)
Radius (Metric)96 mm
Radius (Metric) Min.26 mm
BiocontainmentYes
Capacity1350 mL
MaterialTitanium
Max. RCF42,913 x g
Max. Speed20,000 rpm
TypeZonal Rotor
Unit SizeEach
Frequently asked questions (FAQs)
How long will the delivery take?
It is location dependent. Delivery times typically range from 2-8 weeks depending on product availability, customization requirements, and your location. Contact your sales representative for current lead times.
What is the warranty on Fiberlite rotors and metal rotors used with LYNX centrifuges?
Metal rotors 10 years, Fiberlite carbon fiber rotors 15 years
Where can I find chemical resistance information for the rotors and accessories?
Chemical resistance charts are included in the user manual and additional information is available through technical support.
How can I know what tubes can be used in my LYNX rotors?
The rotor guide of LYNX centrifuges is available on the product page under #documents