TX-750 4 x 750 mL Swinging Bucket Rotor
TX-750 4 x 750 mL Swinging Bucket Rotor
TX-750 4 x 750 mL Swinging Bucket Rotor
TX-750 4 x 750 mL Swinging Bucket Rotor
TX-750 4 x 750 mL Swinging Bucket Rotor
Thermo Scientific™
TX-750 4 x 750 mL Swinging Bucket Rotor
Increase lab productivity with the 750 mL swinging bucket rotor, delivering maximum flexibility at 4700 rpm.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoCapacityTypeFor Use With (Equipment)
75003609Not ApplicableBucket LidRound Buckets
750031804 x 750 mLSwinging Bucket RotorSorvall Legend XT/XF, Sorvall ST 40, Multifuge X3/X3F, Megafuge 40, SL 40/40F
75003608750 mLSwinging Bucket RotorSorvall Legend XT/XF, Sorvall ST 40, Multifuge X3/X3F, Megafuge 40, SL 40/40F
75003610Not ApplicableO-ringClickSeal Biocontainment Lids
75003614250 mLSwinging Bucket RotorSorvall Legend XT/XF, Sorvall ST 40, Multifuge X3/X3F, Megafuge 40, SL 40/40F
75003615Not ApplicableBucket LidRectangular Buckets
750036174 x 750 mLMicroplate/Flask CarrierSorvall Legend XT/XF, Sorvall ST 40, Multifuge X3/X3F, Megafuge 40, SL 40/40F
750037954 x 750 mLMicroplate/Flask CarrierSorvall Legend XT/XF, Sorvall ST 40, Multifuge X3/X3F, Megafuge 40, SL 40/40F
Número do catálogo 75003609
Preço (BRL)
-
Capacity:
Not Applicable
Type:
Bucket Lid
For Use With (Equipment):
Round Buckets
Increase lab productivity with the Thermo Scientific™ TX-750 4 x 750mL Swinging Bucket Rotor, delivering maximum flexibility at 4700rpm.
Ordering Information:
Buckets, lids and microplate carriers are sold separately; order TX-750 rotor body (75003180) and components to complete assembly. For replacement O-rings for lids (set of four) order 75003610. Tube adapters are also available separately. Contact your sales representative for information.
Includes:
Rotor manual
Compatible with:
Thermo Scientific Sorvall™ ST40/ST40R, XT/XTR/XF/XFR, Megafuge™ 40/40R, Multifuge™ X3/X3R, and Thermo Scientific SL40/40R
Recommended for:
Cell culture, blood separation, body fluids processing, microbiology, proteomics, genomics and molecular biology
Especificações
Certifications/ComplianceBiocontainment certified by CAMR™, HPA, Porton Down, U.K.
DescriptionClickSeal Biocontainment Lids
For Use With (Equipment)Round Buckets
QuantitySet of 4
BiocontainmentYes
CapacityNot Applicable
TypeBucket Lid
Unit SizePack of 4