Thermo Scientific™
TX-750 4 × 750mL Swinging Bucket Rotor
Related applications:
Centrifuges
Increase lab productivity with the Thermo Scientific™ TX-750 4 × 750mL Swinging Bucket Rotor, delivering maximum flexibility at 4700rpm.
Back to top
Ordering Information:
Buckets, lids and microplate carriers are sold separately; order TX-750 rotor body (75003180) and components to complete assembly. For replacement O-rings for lids (set of four) order 75003610. Tube adapters are also available separately. Contact your sales representative for information.
Includes:
Rotor manual
Compatible with:
Thermo Scientific Sorvall™ ST40/ST40R, XT/XTR/XF/XFR, Heraeus™ Megafuge™ 40/40R, Multifuge™ X3/X3R, and Thermo Scientific SL40/40R
Recommended for:
Cell culture, blood separation, body fluids processing, microbiology, proteomics, genomics and molecular biology