BIOShield™ 720 Rotor Tube Adapters
BIOShield™ 720 Rotor Tube Adapters
BIOShield™ 720 Rotor Tube Adapters
Thermo Scientific™
BIOShield™ 720 Rotor Tube Adapters
Enhance your rotor's versatility with Thermo Scientific™ BIOShield™ 720 Tube Adapters, which support a wide assortment of tube sizes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity
7500367750 mL Conical Tube
7500367815 mL Conical Tube
7500370110 mL Blood Collection Tube
7500382015 mL Starstedt Tube
750038215/7 mL Blood Collection Tube
Catalog number 75003677
Price (MXN)
-
Capacity:
50 mL Conical Tube
Enhance your rotor's versatility with Thermo Scientific™ BIOShield™ 720 Tube Adapters, which support a wide assortment of tube sizes. These adapters are for use with the BIOShield 720 High Speed Swinging-Bucket Rotor.
  • Maximum tube capacity: 4 × 150mL
  • Maximum O.D. × L: 50 × 122mm
Specifications
Tube Size50 mL
Tube TypeConical
Capacity50 mL Conical Tube
TypeRotor Tube Adapter
Unit SizeEach