TX-400 Rotor Adapters
Thermo Scientific™
TX-400 Rotor Adapters
Use these rotor adapters with round buckets, which enable rotor to accept bottles up to 250 mL and tubes up to 100 mL.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity
7500368076 x 5/7 mL Blood Collection Tube
7500368156 x 10/15 mL Blood Collection Tube
7500368215 mL Conical Tube
7500368350 mL Conical Tube
750037001.5/2.0 mL Conical Microtube
7500370330/25 mL DIN Round/Flat-bottom
7500370415 mL Round-bottom Sarstedt
7500370625 mL Universal
75003708100 mL Round-bottom, Open-tube
75003788175, 200, 225, 250 mL Bottle
750037933 mL RIA or Round-bottom without Cap
7500379415 mL Blood Collection Tube
7500379814 mL Conical Urine Tube
Catalog number 75003680
Price (MXN)
-
Capacity:
76 x 5/7 mL Blood Collection Tube
Use Thermo Scientific™ TX-400 Adapters with round buckets in the TX-400 Rotor.
The adapters enable rotor to accept bottles up to 250mL and tubes to 100mL.
  • Maximum adapter capacity: 4 × 250mL (4 × 400mL with 400mL polypropylene bio-bottles)
  • Maximum adapter O.D. × L: 62 × 125mm (80 × 125mm with 400mL bio-bottles)
Specifications
For Use With (Equipment)TX-400 Rotor
Max. Tube Diameter13 mm
Max. Tube Length110 mm
No. per Pack4
Tube Type5/7 mL Blood Collection Tube
Capacity76 x 5/7 mL Blood Collection Tube
TypeRound Bucket Adapter
Unit SizeCase of