BIOLiner™ Rotor Bucket Adapters
Thermo Scientific™
BIOLiner™ Rotor Bucket Adapters
Combine these rotor bucket adapters with the BIOLiner Rotor Bucket for a complete system.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity
75003691250 mL Corning Conical Bottles
750036715/7 mL Blood Collection Tube
7500367210 mL Blood Collection Tube
7500367315 mL Conical Tube
7500367440 x 50 mL Conical Tubes
75003675250 mL Nalgene Oak Ridge Bottles
75003692750 mL Flat-bottom Bottle w/Round Adapters
Catalog number 75003691
Price (EUR)
-
Capacity:
250 mL Corning Conical Bottles
Combine Thermo Scientific™ BIOLiner™ Rotor Bucket Adapters with the BIOLiner Rotor Bucket for a complete system.
Ordering Information:
Individual adapters and bottle/adapter packages are offered.
  • Capacity range: 3L to 980mL
  • Maximum tube dimensions (O.D. x L): 98 × 124mm
Includes:
75-003-692 includes 750mL flat-bottom bottle with all round adapters for the TX-750 round bucket and four 500mL Corning™ conical bottle adapters.
Specifications
No. per Pack4
Tube Type250 mL Corning Conical Bottle
Capacity250 mL Corning Conical Bottles
TypeRotor Bucket Adapter
Unit SizeEach