TX-750 Round Bucket Adapters
Thermo Scientific™
TX-750 Round Bucket Adapters
Add functionality to your rotor with round bucket adapters, which provide support for round-bottom and conical tubes and bottles, microtubes, and blood collection tubes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity
7500371984 x 15 mL Blood Collection Tube
7500363850 mL Conical Tube
7500363915 mL Conical Tube
75003710250 mL Bottle
75003713100 mL Round-bottom Open-top Tube
7500371550 mL Round-bottom Tube
7500371625 mL Universal Conical/Skirted Tube
7500371814 mL Conical Urine Tube
750037235/7 mL Blood Collection Tube
750037245 mL RIA/Round-bottom Tube (w/o cap)
750037325/7 mL Round-bottom Tube (w/o cap) w/Decanting Aid
750037331.5/2 mL Conical Microtube
75003792250 mL Corning Conical Bottle
Catalog number 75003719
Price (MXN)
-
Capacity:
84 x 15 mL Blood Collection Tube
Add functionality to your rotor with Thermo Scientific™ TX-750 Round Bucket Adapters, which provide support for round-bottom and conical tubes and bottles, microtubes and blood collection tubes.
Adapters will hold round-bottom and conical tubes and bottles, microtubes and blood collection tubes.
Specifications
For Use With (Equipment)Round-Bottom and Conical Tube and Bottle, Microtube and Blood Collection Tube
Tube Type15 mL Blood Collection Tube
Capacity84 x 15 mL Blood Collection Tube
TypeRound Bucket Adapter
Unit SizeEach