Centrifugation Polypropylene Bio-Bottles
Thermo Scientific™
Centrifugation Polypropylene Bio-Bottles
Use these polypropylene screw cap closure bio-bottles for compatibility with round-bucket centrifuge systems.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호용량(미터법)
75003800180 mL
75007585400 mL
카탈로그 번호 75003800
제품 가격(KRW)
700,000
Each
용량(미터법):
180 mL
제품 가격(KRW)
700,000
Each
Use these polypropylene screw cap closure bio-bottles for centrifugation compatibility with round buckets as in the Thermo Scientific™ TX-400 Rotor.
  • With screw cap
  • The 180mL Bio-Bottles (75-003-800) are compatible with Round Buckets for Thermo Scientific TX-200 Rotor (75-003-569)
  • The 400mL Bio-Bottles (75-007-585) are compatible with Round Buckets for Thermo Scientific TX-400 Rotor (75-003-655)
사양
용량12 x 180 mL
마개 재질Polypropylene
마개 유형Screw Cap
용도(장비)Round Bucket for Thermo Scientific TX-200 Rotor
용량(미터법)180 mL
색상Clear
물질Polypropylene
제품 유형Centrifuge Bottle
형상Round
Unit SizeEach