Buckets for Thermo Scientific™ HAEMAFlex™ Swinging Bucket Rotors
Buckets for Thermo Scientific™ HAEMAFlex™ Swinging Bucket Rotors
Buckets for Thermo Scientific™ HAEMAFlex™ Swinging Bucket Rotors
Buckets for Thermo Scientific™ HAEMAFlex™ Swinging Bucket Rotors
Buckets for Thermo Scientific™ HAEMAFlex™ Swinging Bucket Rotors
Buckets for Thermo Scientific™ HAEMAFlex™ Swinging Bucket Rotors
Thermo Scientific™
Buckets for Thermo Scientific™ HAEMAFlex™ Swinging Bucket Rotors
Expand individual bucket capacity or maximize total blood bags per spin with choice of buckets for Thermo Scientific™ HAEMAFlex™ blood banking rotors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Description
75003834HAEMAFlex 6/8 Swing Bucket RotorOval Single Bucket
75003846HAEMAFlex 12/16 Swing Bucket RotorDouble Bucket
75003836HAEMAFlex 6/8 Swing Bucket RotorRound Bucket
75003835HAEMAFlex 6/8 Swing Bucket RotorSingle with Filter Pack
75003865HAEMAFlex 12/16 Swing Bucket RotorDouble with Dura-Coat
75003863HAEMAFlex 6/8 Swing Bucket RotorOval with Dura-Coat
75003864HAEMAFlex 6/8 Swing Bucket RotorRound with Dura-Coat
Catalog number 75003834
Price (BRL)
-
For Use With (Equipment):
HAEMAFlex 6/8 Swing Bucket Rotor
Description:
Oval Single Bucket
Thermo Scientific blood banking centrifuges offer the flexibility to select higher capacity rotors or match existing workflows, with a choice of Thermo Scientific™ HAEMAFlex™ rotors and bucket systems.
Address your lab’s regular- to medium-throughput blood processing needs with the HAEMAFlex 6 swinging bucket rotor with 6 x 550mL blood bag capacity or, utilizing the same bucket system, upgrade to 8 x 550mL bags with the HAEMAFlex 8 swinging bucket rotor.
For high-throughput blood processing needs, the HAEMAFlex 12 swinging bucket rotor provides 12 x 500mL capacity or upgrade your centrifuge capacity to 16 x 500mL blood bags with the HAEMAFlex 16 rotor and the same bucket selections.
Upgrade to Thermo Scientific™ Dura-Coat™ buckets with nickel-coating to improve protection against corrosion due to moisture, chemicals or alkaline solutions.
Specifications
Capacity16 x 500 mL
DescriptionOval Single Bucket
For Use With (Equipment)HAEMAFlex 6/8 Swing Bucket Rotor
TypeSingle Blood Bag Bucket
Unit SizeEach