Accessories for Thermo Scientific™ HAEMAFlex™ Rotors
Actual product may vary
Accessories for Thermo Scientific™ HAEMAFlex™ Rotors
Accessories for Thermo Scientific™ HAEMAFlex™ Rotors
Thermo Scientific™
Accessories for Thermo Scientific™ HAEMAFlex™ Rotors
Complete your rotor system with accessories for Thermo Scientific™ HAEMAFlex™ rotors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Type
75003844HAEMAFlex Swinging Bucket RotorVolume Compensation Plate
75003832Double Blood Bag AdaptersLiner Stand
75003833Single Blood Bag AdaptersLiner Stand
75003843HAEMAFlex Swinging Bucket RotorBlood Bag Spacer
75003845HAEMAFlex Swinging Bucket RotorBalance Plate
75003866HAEMAFlex Swinging Bucket RotorDummy Weight
Catalog number 75003844
Price (TWD)
-
For Use With (Equipment):
HAEMAFlex Swinging Bucket Rotor
Type:
Volume Compensation Plate
Thermo Scientific blood banking centrifuges offer the flexibility to select higher capacity rotors or match existing workflows, with a choice of Thermo Scientific™ HAEMAFlex™ rotors and bucket systems.
Accessories designed to complete your HAEMAFlex rotor system.
Specifications
MaterialRubber
QuantitySet of 12
For Use With (Equipment)HAEMAFlex Swinging Bucket Rotor
TypeVolume Compensation Plate
Unit SizeEach