Search
Search

| Catalog Number | Product Type | Dimensions (L x W) |
|---|---|---|
| 75004117 | Low Adhesion Mat | 280 x 170 mm |
| 75004111 | Low Adhesion Mat | 200 x 200 mm |
| 75004126 | High Adhesion Mat | 200 x 200 mm |
| 75004127 | High Adhesion Mat | 280 x 170 mm |
| 300349 | Adhesive Mat | 230 x 230 mm |
Mix and match Thermo Scientific™ Solaris™ shaker accessories to suit individual needs in the laboratory. Adhesive mats are compatible with all Thermo Scientific™ Solaris and MaxQ platforms and can be cut to size.
Mix and match Thermo Scientific™ Solaris™ shaker accessories to suit individual needs in the laboratory. Adhesive mats are compatible with all Thermo Scientific™ Solaris and MaxQ platforms.
* The maximum speed depends on the type of vessel, as distinguished by size, shape, bottom shape, material, surface roughness and cleanliness. You must evaluate these parameters to determine the optimum choice of mat for your application before using the shaker. Additional guidelines on cleaning, installation, and use can be found in the Thermo Scientific™ Solaris™ Shaker Series Instruction Manual