Adapters for TX-1000 Swinging Bucket Rotor
Thermo Scientific™

Adapters for TX-1000 Swinging Bucket Rotor

An assortment of adapters for outstanding flexibility
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호용량용도(장비)
750042534 x 500 mL500mL Nalgene Bottle
750073014 x 1000 mL1000mL Bio-Bottle
7500425248 x 50 mL50mL Thermo Scientific Nalgene Oak Ridge Tube
750053924 x 250 mL250mL Conical Bottle or 200mL Thermo Scientific Nunc Bottle or 175mL Thermo Scientific Nalgene Conical Bottle
750073024 x 500 mL500mL Corning Bottle
750073034 x 6 mL Standard or 4 x 2 mL Deep-WellMicroplate carriers
750073044 x 750 mL750 mL Polypropylene Wide-Mouth Bio-Bottle and any TX-750 Round adapter
750073058 x 250 mL250mL Thermo Scientific Nalgene Bottle
7500730696 x 15 mL15mL Conical Tube
카탈로그 번호 75004253
제품 가격(KRW)
1,357,000
온라인 행사
Ends: 30-Sep-2026
1,596,000
할인액 239,000 (15%)
Each
용량:
4 x 500 mL
용도(장비):
500mL Nalgene Bottle
제품 가격(KRW)
1,357,000
온라인 행사
Ends: 30-Sep-2026
1,596,000
할인액 239,000 (15%)
Each
An assortment of adapters for outstanding flexibility
사양
용도(장비)500mL Nalgene Bottle
최대 튜브 직경70 mm
최대 튜브 길이160 mm
튜브 유형500 mL Nalgene Bottle
용량4 x 500 mL
유형Rotor Adapter
Unit SizeEach