Thermo Scientific™
Megafuge™ 16 Centrifuge Series
Related applications:
Centrifuges
Process everyday samples with the Thermo Scientific™ Megafuge™ 16 Centrifuge Series, which offers exceptional capacity, ergonomics and value.Perfect for clinical protocols, cell culture applications and microplate processing, the refrigerated version is designed for handling temperature-sensitive sample processing between -10° and +40°C.
Back to top
Easy To Use:
  • Intuitive controls simplify operation and provide detailed information on a wide range of operating parameters and processing status for optimal performance.
  • Motorized lid latch enables one finger downward motion for easiest closing and locking of centrifuge lid, even on tall benches.
Thermo Scientific™ Auto-Lock™ III Rotor System:
  • Secure locking system for easy push-button installation and exchange of rotors
  • Saves time switching between applications
  • Easy access to rotor chamber for quick cleaning, a healthier working environment and longer unit life
Thermo Scientific™ ClickSeal™ Bucket Sealing System:
  • Biocontainment solution that seals with a snap; eliminates screw caps and complicated clips
  • Glove-friendly, one-handed open/close capability
  • Certified by CAMR™ in Porton Down, UK
Thermo Scientific™ SMARTSpin™ Technology:
  • Advanced rotor management system maximizes acceleration, braking and residual load imbalance for each rotor and bucket option
  • Optimizes safety and improves separations
Space-Savings Design:
  • Compact dimensions optimize work area.
  • Ergonomic height and curved design ensure easy sample loading and unloading and easy cleaning.
Outstanding Energy Efficiency:
  • Up to 40% energy consumption savings on industry standard protocols, such as standard blood separation or conical tubes processing
Ventilated Megafuge 16:
  • Dimensions: 23.8L × 17.3W × 14.2 in. H [34.3 in.H open] (60.5 × 44 × 36cm [87cm open])
  • Net weight: 126.8 lb. (57.5kg)
Refrigerated Megafuge 16R:
  • CFC-free refrigeration system
  • Temperature range: -10° to +40°C
  • Pre-cooling function with direct button
  • 23.8L × 24.5W × 14.2 in. H [34.3 in.H open] (60.5 × 62.3 × 36cm [87cm open])
  • Net weight: 201.7 lb. (91.5kg)
Ordering Information:
A wide range of rotors are available for your daily routine processing requirements. Maximize performance with lighter-weight Thermo Scientific Fiberlite™ carbon fiber rotors that offer versatility, speed and robust, corrosion-free design. All rotors and adapters are sold separately.
Includes:
  • Smartspin Technology
  • Rotors, adapters and tubes must be ordered separately to accommodate your application.
Recommended for:
  • Cell Culture
  • Blood Processing
  • Microplates