Sorvall™ ST 8 Small Benchtop Centrifuge
Thermo Scientific™

Sorvall™ ST 8 Small Benchtop Centrifuge

Meet space, clinical and research protocal needs with the compact and innovative Thermo Scientific™ Sorvall™ ST 8 Small Benchtop Centrifuge. Rotors and buckets are sold separately.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberElectrical RequirementsRefrigeratedDescription
75007205100 V 50/60 HzYesSorvall ST 8R Small Benchtop Centrifuge
75007200120 V 60 HzNoSorvall ST 8 Small Benchtop Centrifuge
75007201230 V 50/60 HzNoSorvall ST 8 Small Benchtop Centrifuge
75007202100 V 50/60 HzNoSorvall ST 8 Small Benchtop Centrifuge
75007203120 V 60 HzYesSorvall ST 8R Small Benchtop Centrifuge
Catalog number 75007205
Price (MXN)
-
Electrical Requirements:
100 V 50/60 Hz
Refrigerated:
Yes
Description:
Sorvall ST 8R Small Benchtop Centrifuge
Meet clinical and research protocol needs with the Thermo Scientific™ Sorvall™ ST 8 Centrifuge Series - available as ventilated and refrigerated models. A floor-standing refrigerated version is also available (Sorvall ST 8 FR). These centrifuges provide exceptional capacity in a compact design with a smart, simple interface and the flexibility to support numerous applications.

Innovative technologies:

  • Secure, push-button Thermo Scientific™ Auto-Lock™ rotor exchange in as little as three seconds delivers trouble-free rotor installation and removal and clear chamber access for cleaning convenience.
  • Biocontainment sealing options, including certified† Thermo Scientific™ ClickSeal™ lids offer glove-friendly, one-handed operation.
    • Maximized swing-out capacity up to 8 x 50mL conical tubes, 24 x 5/7mL blood tubes, microplates and microtubes, all in one versatile centrifuge
    • Fixed angle flexibility to spin 50mL conical tubes at over 12,000 x g or a wide range of microtubes at speeds up to 30,279 x g

User-friendly performance:

  • Low noise levels, including just 52 dBA at performance of 17,850rpm/30,279 x g (with Sorvall ST 8R centrifuge), to ensure a peaceful laboratory
  • One-touch operation with pre-saved protocols
  • Highly visible backlit display offers easy reading of parameters across the lab
  • Glove- and detergent-friendly user interface
  • Multilingual instructions offered in English, Dutch, French, German, Italian, Russian and Spanish on programming, run conditions, alerts and service messages
  • Centrifuge conforms to the latest clinical and safety standards such as UL, CE and IVD

Clinical applications:

  • Runs up to 24 x 5/7mL blood tubes at a time in swinging bucket configuration
  • Conforms to the latest clinical and safety standards

Research applications:

  • 8 x 50mL swinging bucket rotor capacity, four microplates or 30 filtration columns for research needs
  • Auto-Lock rotor exchange for application flexibility—go from 50mL tube to a microplate to microtubes effortlessly

†Biocontainment certification by the Public Health England, Porton Down, UK.

NOTE: Rotors and buckets must be purchased separately.
Specifications
Certifications/ComplianceIEC 61010-1, IEC 61010-2-020, IEC 61010-2-101, EN 61326-1; UL Listed / CE marked /IVD compliant / Biocontainment certified by the Public Health England, Porton Down, UK
Controller TypeMicroprocessor
DescriptionSorvall ST 8R Small Benchtop Centrifuge
Dimensions (HWD)12.6 x 18.1 x 26.4 in. (32 x 46 x 67 cm)
Drive SystemDirect Brushless Induction
Electrical Requirements100 V 50/60 Hz
Frequency50/60 Hz
Height (English) Exterior12.6 in.
Height (English) Exterior Lid Open27.5 in.
Height (Metric) Exterior32 cm
Height (Metric) Exterior Lid Open70 cm
IncludesST 8 Centrifuge. (Rotors and buckets are sold separately.)
Length (English) Exterior26.4 in.
Length (Metric) Exterior67 cm
Net Weight156 lb. (71 kg)
Noise Level<52 dB(A) (with MicroClick 24 x 2 rotor); <56 dB(A) with all rotors
Profile (Acceleration/Braking)2 (standard and soft)
Program Storage3 direct program buttons, plus 96 additional programs accessible via folder, all with alphanumeric program naming
RotorTX-150
Rotor IncludedNo
Run Time99 hr. 59 min., continuous
Safety FeaturesAuto-Lock rotor exchange system, SMARTSpin imbalance detection system, certified ClickSeal biocontainment lids
Temperature Range (Metric)-10°C to +40°C
Wattage750 w
Width (English) Exterior18.1 in.
Width (Metric) Exterior46 cm
Capacity4 x 145 mL (TX-150), 6 x 50 mL (HIGHConic III)
Max. RCF3,260 x g (TX-150 Rotor); 30,279 x g (MicroClick 24 x 2 Rotor)
Max. Speed4,500 rpm (TX-150 rotor), 17,850 rpm (MicroClick 24 x 2 rotor)
Product LineSorvall
RefrigeratedYes
Unit SizeEach