Sorvall™ Lid for Primo™ Round Buckets
Thermo Scientific™
Sorvall™ Lid for Primo™ Round Buckets
Provides biocontainment protection
Have Questions?
Catalog NumberFor Use With (Equipment)
75007598Thermo Scientific Sorvall Swing Bucket
Catalog number 75007598
Price (MXN)
-
For Use With (Equipment):
Thermo Scientific Sorvall Swing Bucket
Provides biocontainment protection
Lids fit Round Buckets (75-007-555) in 4-Place Swinging Bucket Rotor (75-007-591). Sold individually; order correct quantity for rotor.
Specifications
For Use With (Equipment)Thermo Scientific Sorvall Swing Bucket
Unit SizeEach