C Series Ventilated Centrifuges
Thermo Scientific™
C Series Ventilated Centrifuges
The Thermo Scientific™ C Series Centrifuges are designed for clinical and research applications, offering exceptional capacity in a compact design.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeVoltage
75008005IVD-MD230 V
75008004IVD-MD120 V
Catalog number 75008005
Price (MXN)
-
Type:
IVD-MD
Voltage:
230 V
The Thermo Scientific™ C Series Centrifuges are designed for clinical and research applications, offering exceptional capacity in a compact design. The centrifuges are available as ventilated or refrigerated models. The refrigerated models feature Thermo Scientific™ GreenCool™ Technology, a next-generation natural refrigerant cooling system for enhanced sustainability. With a fixed-angle rotor RCF (Relative Centrifugal Force) up to 30,279 x g and a swinging bucket rotor RCF up to 3,260 x g, the C Series Centrifuges are capable of handling even the toughest separations. The maximum capacity of the fixed-angle rotors is 6 x 50 mL, while the swinging bucket rotors can accommodate up to 4 x 145 mL. These centrifuges combine a compact design with an intuitive interface and the flexibility to support numerous applications.
  • High capacity in a compact design
  • GreenCool Technology (refrigerated models)
  • Thermo Scientific™ Auto-Lock™ Rotor Exchange
  • Thermo Scientific™ ClickSeal™ Biocontainment Lids
  • Intuitive user interface
  • Wide range of rotors for application flexibility
Specifications
Certifications/ComplianceUL Listed, CE marked, IVD compliant, Certified biocontainment, IEC 61010-1, IEC 61010-2-020, IEC 61010-2-101, EN 61326-1
Dimensions (L x W x H) Exterior48 x 37 x 31 cm (18.9 x 14.6 x 12.2 in)
DisplayKeypad
For Use With (Application)Clinical and research applications
IncludesCentrifuge
Net Weight35 kg (77 Ib)
Noise Level<60 dB (A) with a TX-150 rotor (at max. speed)<65,5 dB (A) with a MicroClick 24x2 rotor (at max. speed)
TypeIVD-MD
VentilatedYes
Voltage230 V
Capacity4 x 145 mL
Max. RCF3,260 x g (TX-150 Rotor); 24,328 x g (MicroClick 24 x 2 Rotor)
Max. Speed16,000 rpm
RefrigeratedNo
Unit SizeEach