Specifications
Certifications/ComplianceUL Listed, CE marked, IVD compliant, Certified biocontainment, IEC 61010-1, IEC 61010-2-020, IEC 61010-2-101, EN 61326-1
Dimensions (L x W x H) Exterior67 x 46 x 32 cm (26.4 x 18.1 x 12.6 in)
Display TypeKeypad
For Use With (Application)Clinical and research applications
IncludesCentrifuge
ModelC1TR: IVD-MD
Net Weight71 kg (156 Ib)
Noise Level<47 dB(A) with TX-150 rotor (conical bucket) <46 dB(A) with MicroClick 24x2 rotor at top speed
VentilatedNo
Voltage120 V
Capacity4 x 145 mL
Max. RCF3,260 x g (TX-150 Rotor); 30,279 x g (MicroClick 24 x 2 Rotor)
Max. Speed17,850 rpm
RefrigeratedYes
Unit SizeEach