Regulatory requirements vary by country; products may not be available in your geographic area.

PrioMab CSFV Erns Monoclonal Antibody 140.1 (2 mg/mL)

Catalog number: 7500980

 Related applications: Animal Health

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Monoclonal antibody rainsed against the Erns protein of the classical swine fever virus (CSFV).
For Veterinary Use Only.

Specifications

Clonality: Monoclonal
Host Species: Mouse
Product Line: PrioMab, Prionics™
Product Size: 1 mg
Reactivity: Swine

Documents

Manuals & protocols