Regulatory requirements vary by country; products may not be available in your geographic area.

CSFV Polyclonal Antibody (2 mg/mL)

Catalog number: 7500989

 Related applications: Animal Health

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Polyclonal antibody that reacts with all pestiviruses: bovine viral diarrhoea virus, border disease virus, and classical swine fever, including the chinese vaccine strain.
For Veterinary Use Only.

Specifications

Clonality: Polyclonal
Host Species: Swine
Product Line: Prionics™
Product Size: 1.5 mL
Reactivity: Swine

Documents

Manuals & protocols