Regulatory requirements vary by country; products may not be available in your geographic area.

Prion Monoclonal Antibody 6H4

Catalog number: 7500996

 Related applications: Animal Health

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Monoclonal anti-PrP 6H4 is a mouse IgG1 subtype antibody. Light chain: k subtype. mAb 6H4 recognizes the sequence DYEDRYYRE of the prion protein (human PrP: amino acids 144-152). The antibody reacts with most known mammalian PrP sequences (human, cattle, sheep, rabbit, mink, a variety of primates, mouse, hamster, and rat). The antibody is used in numerous scientific publications and applied in several methods, including Western Blot, ELISA, immunohistochemistry (IHC), immunoprecipitation, PET blot, and FACS analysis. The antibody is also the principal component of all of Prionics®-Check TSE kits.
For Veterinary Use Only.

Specifications

Clonality: Monoclonal
Host Species: Mouse
Product Line: Prionics™
Product Size: 100 µg
Reactivity: Bovine, Hamster, Mink, Mouse, Primate, Rabbit, Rat, Sheep

Documents

Manuals & protocols