Regulatory requirements vary by country; products may not be available in your geographic area.

Prion Monoclonal Antibody 34C9

Catalog number: 7500998

 Related applications: Animal Health

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Monoclonal anti-PrP 34C9 is a mouse IgG2b subtype antibody. Light chain: k subtype. mAB 34C9 recognizes the sequence LIHFG in the bovine prion protein (corresponding to position 138-142 with numbering according to the human PrP). This sequence is conserved in bovine, rabbit, pig, and mule deer PrP sequences, but not in human, mouse, or hamster PrP. mAb 34C9 is suited for Western blotting.
For Veterinary Use Only.

Specifications

Clonality: Monoclonal
Host Species: Mouse
Product Line: Prionics™
Product Size: 100 µg
Reactivity: Bovine, Mule, Rabbit, Swine

Documents

Manuals & protocols