Regulatory requirements vary by country; products may not be available in your geographic area.

Prion Monoclonal Antibody 2E11

Catalog number: 7501001

 Related applications: Animal Health

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Monoclonal anti-PrP 2E11 is a mouse IgG2a subtype monoclonal antibody. The antibody reacts with bovine, sheep (only Q171 alleles), deer, and elk prion protein.
For Veterinary Use Only.

Specifications

Clonality: Monoclonal
Host Species: Mouse
Product Line: Prionics™
Product Size: 1 mg
Reactivity: Bovine, Deer, Moose, Sheep

Documents

Manuals & protocols