Thermo Scientific™
Multifuge X4 and X4R Pro Centrifuges
Catalog number: 75016081
Thermo Scientific™
Multifuge X4 and X4R Pro Centrifuges
Catalog number: 75016081
Thermo Scientific™ Multifuge™ X4 and X4R General Purpose Pro Centrifuges provide high performance and versatility with an easy-to-use, full-color touch screen user interface. The centrifuges make programming easy for quick results and allow easy access to and tracking of all runs, rotor life and centrifuge health. With up to 4L capacity, they allow spinning 196 blood tubes, 40x 50mL conical tubes and 96x 15mL conical tubes. Maximum versatility is achieved with 18 available rotors to choose from and Thermo Scientific™ Auto-Lock™ Rotor Exchange to quickly change between applications.
Available for purchase individually or in convenient application-based packages:
Cell Culture packages include the centrifuge, rotor, ClickSeal biocontainment lids and a set of four 15 mL and 50 mL conical tube adapters.
Blood Tube packages include the centrifuge, rotor, ClickSeal biocontainment lids and a set of four 5/7 mL and 9/10 mL blood tube adapters.
Catalog Number
75016081
Unit Size
Each
Description
Multifuge X4R Pro-MD, 220V-240V 50Hz/230V 60Hz, TX-750 Blood Tube Package
Voltage
220/240 V
Rotor Included
Yes
Price (USD)
Availability
-
Full specifications
Quantity
-
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller TypeMicroprocessor
DescriptionMultifuge X4R Pro-MD, 220V-240V 50Hz/230V 60Hz, TX-750 Blood Tube Package
Capacity148 x 5/7 mL Blood Tubes, 84 x 9/10 mL Blood Tubes
Max. RCF4,816 x g (with TX-750 rotor)
Max. Speed4,700 rpm (with TX-750 rotor)
Product LineMultifuge
RefrigeratedYes
DisplayFull Color Touch Screen Interface
Drive SystemDirect, Brushless Induction Low Profile
Green FeaturesLess waste, Energy efficient
IncludesPrinted English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
MaterialSee individual centrifuge, rotor and accessory data
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
RotorBlood Tube Package Includes TX-750 with Buckets, ClickSeal Biocontainment Lids, 5/7 and 9/10 mL Blood Tube Adapters
Rotor IncludedYes
Run TimeUp to 99 hr. 59 min. 59 sec.
StandardsISO 14971, ISO 13485, ISO 9001
Temperature Range-10°C to +40°C, ±2°C
TimerOption to count up or down
TypeBenchtop Centrifuge
VentilatedYes
WarrantyCentrifuge: 5 Year Limited, Rotor: See Manual
Net WeightSee individual components of package
Voltage220/240 V
Frequency50/60 Hz
Laboratory/Cleanroom UseCleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Multifuge X4 Pro-MD 208-240V 50/60Hz
Voltage
208/240 V
Rotor Included
No
Price (USD)
Availability
-
Full specifications
Quantity
-
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller TypeMicroprocessor
DescriptionMultifuge X4 Pro-MD 208-240V 50/60Hz
Capacity4 x 6 Microplates
Max. RCF7,197 x g Swing-Out with H-Flex HS4 rotor
Product LineMultifuge
RefrigeratedNo
DisplayFull Color Touch Screen Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesEnergy efficient, less waste
IncludesPrinted English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
MaterialFront, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Noise Level<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor18 Rotor options available
Rotor IncludedNo
Run TimeUp to 99 hr. 59 min. 59 sec.
StandardsISO 14971, ISO 13485, ISO 9001
TimerOption to count up or down
TypeBenchtop Centrifuge
VentilatedYes
Warranty5 Year Limited
Dimensions (L x W x H) Exterior690 x 566 x 362 mm
Length (English) Exterior27.2 in.
Length (Metric) Exterior690 mm
Width (English) Exterior22.3 in.
Width (Metric) Exterior566 mm
Height (English) Exterior14.2 in.
Height (English) Exterior Lid Open33.9 in.
Height (Metric) Exterior362 mm
Height (Metric) Exterior Lid Open861 mm
Depth (English)27.2 in.
Depth (Metric)690 mm
Net Weight89 kg
Voltage208/240 V
Wattage1600 w
Frequency50/60 Hz
Laboratory/Cleanroom UseCleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Multifuge X4 Pro-MD 120V
Voltage
120 V
Rotor Included
No
Price (USD)
Price: 14,400.00
Your price:
Availability
-
Full specifications
Quantity
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller TypeMicroprocessor
DescriptionMultifuge X4 Pro-MD 120V
Capacity4 x 6 Microplates
Max. RCF7,197 x g Swing-Out with H-Flex HS4 rotor
Product LineMultifuge
RefrigeratedNo
DisplayFull Color Touch Screen Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesEnergy efficient, less waste
IncludesPrinted English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
MaterialFront, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Noise Level<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor18 Rotor options available
Rotor IncludedNo
Run TimeUp to 99 hr. 59 min. 59 sec.
StandardsISO 14971, ISO 13485, ISO 9001
TimerOption to count up or down
TypeBenchtop Centrifuge
VentilatedYes
Warranty5 Year Limited
Dimensions (L x W x H) Exterior690 x 566 x 362 mm
Length (English) Exterior27.2 in.
Length (Metric) Exterior690 mm
Width (English) Exterior22.3 in.
Width (Metric) Exterior566 mm
Height (English) Exterior14.2 in.
Height (English) Exterior Lid Open33.9 in.
Height (Metric) Exterior362 mm
Height (Metric) Exterior Lid Open861 mm
Depth (English)27.2 in.
Depth (Metric)690 mm
Net Weight89 kg
Voltage120 V
Wattage1300 w
Frequency50/60 Hz
Laboratory/Cleanroom UseCleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Multifuge X4R Pro-MD 220-240V 50Hz / 230V 60Hz
Voltage
220V-240V / 230V
Rotor Included
No
Price (USD)
Availability
-
Full specifications
Quantity
-
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller TypeMicroprocessor
DescriptionMultifuge X4R Pro-MD 220-240V 50Hz / 230V 60Hz
Capacity4 x 6 Microplates
Max. RCF7,197 x g Swing-Out with H-Flex HS4 rotor
Product LineMultifuge
RefrigeratedYes
DisplayFull Color Touch Screen Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesEnergy efficient, less waste
IncludesPrinted English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
MaterialFront, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Noise Level<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor18 Rotor options available
Rotor IncludedNo
Run TimeUp to 99 hr. 59 min. 59 sec.
StandardsISO 14971, ISO 13485, ISO 9001
Temperature Range-10°C to +40°C, ±2°C
TimerOption to count up or down
TypeBenchtop Centrifuge
VentilatedNo
Warranty5 Year Limited
Dimensions (L x W x H) Exterior690 x 746 x 361 mm
Length (English) Exterior27.2 in.
Length (Metric) Exterior690 mm
Width (English) Exterior29.4 in.
Width (Metric) Exterior746 mm
Height (English) Exterior14.2 in.
Height (English) Exterior Lid Open33.9 in.
Height (Metric) Exterior361 mm
Height (Metric) Exterior Lid Open861 mm
Depth (English)27.2 in.
Depth (Metric)690 mm
Net Weight120 kg
Voltage220V-240V / 230V
Wattage1950 w
Frequency50/60 Hz
Laboratory/Cleanroom UseCleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Multifuge X4R Pro-MD 120V
Voltage
120 V
Rotor Included
No
Price (USD)
Price: 19,300.00
Your price:
Availability
-
Full specifications
Quantity
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller TypeMicroprocessor
DescriptionMultifuge X4R Pro-MD 120V
Capacity4 x 6 Microplates
Max. RCF7,197 x g Swing-Out with H-Flex HS4 rotor
Product LineMultifuge
RefrigeratedYes
DisplayFull Color Touch Screen Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesEnergy efficient, less waste
IncludesPrinted English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
MaterialFront, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Noise Level<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor18 Rotor options available
Rotor IncludedNo
Run TimeUp to 99 hr. 59 min. 59 sec.
StandardsISO 14971, ISO 13485, ISO 9001
Temperature Range-10°C to +40°C, ±2°C
TimerOption to count up or down
TypeBenchtop Centrifuge
VentilatedNo
Warranty5 Year Limited
Dimensions (L x W x H) Exterior690 x 746 x 361 mm
Length (English) Exterior27.2 in.
Length (Metric) Exterior690 mm
Width (English) Exterior29.4 in.
Width (Metric) Exterior746 mm
Height (English) Exterior14.2 in.
Height (English) Exterior Lid Open33.9 in.
Height (Metric) Exterior361 mm
Height (Metric) Exterior Lid Open861 mm
Depth (English)27.2 in.
Depth (Metric)690 mm
Net Weight120 kg
Voltage120 V
Wattage1450 w
Frequency50/60 Hz
Laboratory/Cleanroom UseCleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Multifuge X4R Pro-MD 220V 60Hz
Voltage
220 V
Rotor Included
No
Price (USD)
Availability
-
Full specifications
Quantity
-
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller TypeMicroprocessor
DescriptionMultifuge X4R Pro-MD 220V 60Hz
Capacity4 x 6 Microplates
Max. RCF7,197 x g Swing-Out with H-Flex HS4 rotor
Product LineMultifuge
RefrigeratedYes
DisplayFull Color Touch Screen Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesEnergy efficient, less waste
IncludesPrinted English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the accroding country supply voltage, declaration of conformity
MaterialFront, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Noise Level<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor18 Rotor options available
Rotor IncludedNo
Run TimeUp to 99 hr. 59 min. 59 sec.
StandardsISO 14971, ISO 13485, ISO 9001
Temperature Range-10°C to +40°C, ±2°C
TimerOption to count up or down
TypeBenchtop Centrifuge
VentilatedNo
Warranty5 Year Limited
Dimensions (L x W x H) Exterior690 x 746 x 361 mm
Length (English) Exterior27.2 in.
Length (Metric) Exterior690 mm
Width (English) Exterior29.4 in.
Width (Metric) Exterior746 mm
Height (English) Exterior14.2 in.
Height (English) Exterior Lid Open33.9 in.
Height (Metric) Exterior361 mm
Height (Metric) Exterior Lid Open861 mm
Depth (English)27.2 in.
Depth (Metric)690 mm
Net Weight120 kg
Voltage220 V
Wattage1950 w
Frequency60 Hz
Laboratory/Cleanroom UseCleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Multifuge X4R Pro 220V 60Hz
Voltage
220 V
Rotor Included
No
Price (USD)
Availability
-
Full specifications
Quantity
-
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller TypeMicroprocessor
DescriptionMultifuge X4R Pro 220V 60Hz
Capacity4 x 6 Microplates
Max. RCF7,197 x g Swing-Out with H-Flex HS4 rotor
Product LineMultifuge
RefrigeratedYes
DisplayFull Color Touch Screen Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesEnergy efficient, less waste
IncludesPrinted English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
MaterialFront, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Noise Level<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor18 Rotor options available
Rotor IncludedNo
Run TimeUp to 99 hr. 59 min. 59 sec.
StandardsISO 14971, ISO 13485, ISO 9001
Temperature Range-10°C to +40°C, ±2°C
TimerOption to count up or down
TypeBenchtop Centrifuge
VentilatedNo
Warranty5 Year Limited
Dimensions (L x W x H) Exterior690 x 746 x 361 mm
Length (English) Exterior27.2 in.
Length (Metric) Exterior690 mm
Width (English) Exterior29.4 in.
Width (Metric) Exterior746 mm
Height (English) Exterior14.2 in.
Height (English) Exterior Lid Open33.9 in.
Height (Metric) Exterior361 mm
Height (Metric) Exterior Lid Open861 mm
Depth (English)27.2 in.
Depth (Metric)690 mm
Net Weight120 kg
Voltage220 V
Wattage1950 w
Frequency60 Hz
Laboratory/Cleanroom UseCleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Multifuge X4 Pro 208-240V 50/60Hz
Voltage
208/240 V
Rotor Included
No
Price (USD)
Availability
-
Full specifications
Quantity
-
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller TypeMicroprocessor
DescriptionMultifuge X4 Pro 208-240V 50/60Hz
Capacity4 x 6 Microplates
Max. RCF7,197 x g Swing-Out with H-Flex HS4 rotor
Product LineMultifuge
RefrigeratedNo
DisplayFull Color Touch Screen Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesEnergy efficient, less waste
IncludesPrinted English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
MaterialFront, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Noise Level<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor18 Rotor options available
Rotor IncludedNo
Run TimeUp to 99 hr. 59 min. 59 sec.
StandardsISO 14971, ISO 13485, ISO 9001
TimerOption to count up or down
TypeBenchtop Centrifuge
VentilatedYes
Warranty5 Year Limited
Dimensions (L x W x H) Exterior690 x 566 x 362 mm
Length (English) Exterior27.2 in.
Length (Metric) Exterior690 mm
Width (English) Exterior22.3 in.
Width (Metric) Exterior566 mm
Height (English) Exterior14.2 in.
Height (English) Exterior Lid Open33.9 in.
Height (Metric) Exterior362 mm
Height (Metric) Exterior Lid Open861 mm
Depth (English)27.2 in.
Depth (Metric)690 mm
Net Weight89 kg
Voltage208/240 V
Wattage1600 w
Frequency50/60 Hz
Laboratory/Cleanroom UseCleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Multifuge X4R Pro 220-240V 50Hz/230V 60Hz
Voltage
220/240 V
Rotor Included
No
Price (USD)
Availability
-
Full specifications
Quantity
-
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller TypeMicroprocessor
DescriptionMultifuge X4R Pro 220-240V 50Hz/230V 60Hz
Capacity4 x 6 Microplates
Max. RCF7,197 x g Swing-Out with H-Flex HS4 rotor
Product LineMultifuge
RefrigeratedYes
DisplayFull Color Touch Screen Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesEnergy efficient, less waste
IncludesPrinted English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
MaterialFront, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Noise Level<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor18 Rotor options available
Rotor IncludedNo
Run TimeUp to 99 hr. 59 min. 59 sec.
StandardsISO 14971, ISO 13485, ISO 9001
Temperature Range-10°C to +40°C, ±2°C
TimerOption to count up or down
TypeBenchtop Centrifuge
VentilatedNo
Warranty5 Year Limited
Dimensions (L x W x H) Exterior690 x 746 x 361 mm
Length (English) Exterior27.2 in.
Length (Metric) Exterior690 mm
Width (English) Exterior29.4 in.
Width (Metric) Exterior746 mm
Height (English) Exterior14.2 in.
Height (English) Exterior Lid Open33.9 in.
Height (Metric) Exterior361 mm
Height (Metric) Exterior Lid Open861 mm
Depth (English)27.2 in.
Depth (Metric)690 mm
Net Weight120 kg
Voltage220/240 V
Wattage1950 w
Frequency50/60 Hz
Laboratory/Cleanroom UseCleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Multifuge X4R Pro-MD, 120V TX-1000 Blood Tube Package
Voltage
120 V
Rotor Included
Yes
Price (USD)
Price: 20,000.00
Your price:
Availability
-
Full specifications
Quantity
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2FCC part 15
Controller TypeMicroprocessor
DescriptionMultifuge X4R Pro-MD, 120V TX-1000 Blood Tube Package
Capacity196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes
Max. RCF4,122 x g (with TX-1000 rotor)
Max. Speed4,200 rpm (with TX-1000 rotor)
Product LineMultifuge
RefrigeratedYes
DisplayFull Color Touch Screen Interface
Drive SystemDirect, Brushless Induction Low Profile
Green FeaturesLess waste, Energy efficient
IncludesPrinted English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
MaterialFront, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
RotorBlood Tube Package Includes TX-1000 with Buckets, ClickSeal Biocontainment lids, 5/7 and 9/10 mL Blood Tube Adapters
Rotor IncludedYes
Run TimeUp to 99 hr. 59 min. 59 sec.
StandardsISO 14971, ISO 13485, ISO 9001
Temperature Range-10°C to +40°C, ±2°C
TimerOption to count up or down
TypeBenchtop Centrifuge
VentilatedNo
WarrantyCentrifuge: 5 Year Limited, Rotor: See Manual
Net WeightSee individual components of package
Voltage120 V
Frequency60 Hz
Laboratory/Cleanroom UseCleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
Unit SizeEach
Showing 10 of 18Show more
Catalog NumberSpecificationsUnit SizeDescriptionVoltageRotor IncludedPrice (USD)AvailabilityQuantity
75016081Full specifications
EachMultifuge X4R Pro-MD, 220V-240V 50Hz/230V 60Hz, TX-750 Blood Tube Package220/240 VYesRequest A Quote--
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller TypeMicroprocessor
DescriptionMultifuge X4R Pro-MD, 220V-240V 50Hz/230V 60Hz, TX-750 Blood Tube Package
Capacity148 x 5/7 mL Blood Tubes, 84 x 9/10 mL Blood Tubes
Max. RCF4,816 x g (with TX-750 rotor)
Max. Speed4,700 rpm (with TX-750 rotor)
Product LineMultifuge
RefrigeratedYes
DisplayFull Color Touch Screen Interface
Drive SystemDirect, Brushless Induction Low Profile
Green FeaturesLess waste, Energy efficient
IncludesPrinted English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
MaterialSee individual centrifuge, rotor and accessory data
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
RotorBlood Tube Package Includes TX-750 with Buckets, ClickSeal Biocontainment Lids, 5/7 and 9/10 mL Blood Tube Adapters
Rotor IncludedYes
Run TimeUp to 99 hr. 59 min. 59 sec.
StandardsISO 14971, ISO 13485, ISO 9001
Temperature Range-10°C to +40°C, ±2°C
TimerOption to count up or down
TypeBenchtop Centrifuge
VentilatedYes
WarrantyCentrifuge: 5 Year Limited, Rotor: See Manual
Net WeightSee individual components of package
Voltage220/240 V
Frequency50/60 Hz
Laboratory/Cleanroom UseCleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
Unit SizeEach
75009500Full specifications
EachMultifuge X4 Pro-MD 208-240V 50/60Hz208/240 VNoRequest A Quote--
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller TypeMicroprocessor
DescriptionMultifuge X4 Pro-MD 208-240V 50/60Hz
Capacity4 x 6 Microplates
Max. RCF7,197 x g Swing-Out with H-Flex HS4 rotor
Product LineMultifuge
RefrigeratedNo
DisplayFull Color Touch Screen Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesEnergy efficient, less waste
IncludesPrinted English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
MaterialFront, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Noise Level<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor18 Rotor options available
Rotor IncludedNo
Run TimeUp to 99 hr. 59 min. 59 sec.
StandardsISO 14971, ISO 13485, ISO 9001
TimerOption to count up or down
TypeBenchtop Centrifuge
VentilatedYes
Warranty5 Year Limited
Dimensions (L x W x H) Exterior690 x 566 x 362 mm
Length (English) Exterior27.2 in.
Length (Metric) Exterior690 mm
Width (English) Exterior22.3 in.
Width (Metric) Exterior566 mm
Height (English) Exterior14.2 in.
Height (English) Exterior Lid Open33.9 in.
Height (Metric) Exterior362 mm
Height (Metric) Exterior Lid Open861 mm
Depth (English)27.2 in.
Depth (Metric)690 mm
Net Weight89 kg
Voltage208/240 V
Wattage1600 w
Frequency50/60 Hz
Laboratory/Cleanroom UseCleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
Unit SizeEach
75009501Full specifications
EachMultifuge X4 Pro-MD 120V120 VNo
Price: 14,400.00
Your price:
-
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller TypeMicroprocessor
DescriptionMultifuge X4 Pro-MD 120V
Capacity4 x 6 Microplates
Max. RCF7,197 x g Swing-Out with H-Flex HS4 rotor
Product LineMultifuge
RefrigeratedNo
DisplayFull Color Touch Screen Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesEnergy efficient, less waste
IncludesPrinted English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
MaterialFront, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Noise Level<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor18 Rotor options available
Rotor IncludedNo
Run TimeUp to 99 hr. 59 min. 59 sec.
StandardsISO 14971, ISO 13485, ISO 9001
TimerOption to count up or down
TypeBenchtop Centrifuge
VentilatedYes
Warranty5 Year Limited
Dimensions (L x W x H) Exterior690 x 566 x 362 mm
Length (English) Exterior27.2 in.
Length (Metric) Exterior690 mm
Width (English) Exterior22.3 in.
Width (Metric) Exterior566 mm
Height (English) Exterior14.2 in.
Height (English) Exterior Lid Open33.9 in.
Height (Metric) Exterior362 mm
Height (Metric) Exterior Lid Open861 mm
Depth (English)27.2 in.
Depth (Metric)690 mm
Net Weight89 kg
Voltage120 V
Wattage1300 w
Frequency50/60 Hz
Laboratory/Cleanroom UseCleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
Unit SizeEach
75009515Full specifications
EachMultifuge X4R Pro-MD 220-240V 50Hz / 230V 60Hz220V-240V / 230VNoRequest A Quote--
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller TypeMicroprocessor
DescriptionMultifuge X4R Pro-MD 220-240V 50Hz / 230V 60Hz
Capacity4 x 6 Microplates
Max. RCF7,197 x g Swing-Out with H-Flex HS4 rotor
Product LineMultifuge
RefrigeratedYes
DisplayFull Color Touch Screen Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesEnergy efficient, less waste
IncludesPrinted English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
MaterialFront, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Noise Level<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor18 Rotor options available
Rotor IncludedNo
Run TimeUp to 99 hr. 59 min. 59 sec.
StandardsISO 14971, ISO 13485, ISO 9001
Temperature Range-10°C to +40°C, ±2°C
TimerOption to count up or down
TypeBenchtop Centrifuge
VentilatedNo
Warranty5 Year Limited
Dimensions (L x W x H) Exterior690 x 746 x 361 mm
Length (English) Exterior27.2 in.
Length (Metric) Exterior690 mm
Width (English) Exterior29.4 in.
Width (Metric) Exterior746 mm
Height (English) Exterior14.2 in.
Height (English) Exterior Lid Open33.9 in.
Height (Metric) Exterior361 mm
Height (Metric) Exterior Lid Open861 mm
Depth (English)27.2 in.
Depth (Metric)690 mm
Net Weight120 kg
Voltage220V-240V / 230V
Wattage1950 w
Frequency50/60 Hz
Laboratory/Cleanroom UseCleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
Unit SizeEach
75009516Full specifications
EachMultifuge X4R Pro-MD 120V120 VNo
Price: 19,300.00
Your price:
-
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller TypeMicroprocessor
DescriptionMultifuge X4R Pro-MD 120V
Capacity4 x 6 Microplates
Max. RCF7,197 x g Swing-Out with H-Flex HS4 rotor
Product LineMultifuge
RefrigeratedYes
DisplayFull Color Touch Screen Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesEnergy efficient, less waste
IncludesPrinted English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
MaterialFront, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Noise Level<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor18 Rotor options available
Rotor IncludedNo
Run TimeUp to 99 hr. 59 min. 59 sec.
StandardsISO 14971, ISO 13485, ISO 9001
Temperature Range-10°C to +40°C, ±2°C
TimerOption to count up or down
TypeBenchtop Centrifuge
VentilatedNo
Warranty5 Year Limited
Dimensions (L x W x H) Exterior690 x 746 x 361 mm
Length (English) Exterior27.2 in.
Length (Metric) Exterior690 mm
Width (English) Exterior29.4 in.
Width (Metric) Exterior746 mm
Height (English) Exterior14.2 in.
Height (English) Exterior Lid Open33.9 in.
Height (Metric) Exterior361 mm
Height (Metric) Exterior Lid Open861 mm
Depth (English)27.2 in.
Depth (Metric)690 mm
Net Weight120 kg
Voltage120 V
Wattage1450 w
Frequency50/60 Hz
Laboratory/Cleanroom UseCleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
Unit SizeEach
75009615Full specifications
EachMultifuge X4R Pro-MD 220V 60Hz220 VNoRequest A Quote--
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller TypeMicroprocessor
DescriptionMultifuge X4R Pro-MD 220V 60Hz
Capacity4 x 6 Microplates
Max. RCF7,197 x g Swing-Out with H-Flex HS4 rotor
Product LineMultifuge
RefrigeratedYes
DisplayFull Color Touch Screen Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesEnergy efficient, less waste
IncludesPrinted English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the accroding country supply voltage, declaration of conformity
MaterialFront, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Noise Level<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor18 Rotor options available
Rotor IncludedNo
Run TimeUp to 99 hr. 59 min. 59 sec.
StandardsISO 14971, ISO 13485, ISO 9001
Temperature Range-10°C to +40°C, ±2°C
TimerOption to count up or down
TypeBenchtop Centrifuge
VentilatedNo
Warranty5 Year Limited
Dimensions (L x W x H) Exterior690 x 746 x 361 mm
Length (English) Exterior27.2 in.
Length (Metric) Exterior690 mm
Width (English) Exterior29.4 in.
Width (Metric) Exterior746 mm
Height (English) Exterior14.2 in.
Height (English) Exterior Lid Open33.9 in.
Height (Metric) Exterior361 mm
Height (Metric) Exterior Lid Open861 mm
Depth (English)27.2 in.
Depth (Metric)690 mm
Net Weight120 kg
Voltage220 V
Wattage1950 w
Frequency60 Hz
Laboratory/Cleanroom UseCleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
Unit SizeEach
75009815Full specifications
EachMultifuge X4R Pro 220V 60Hz220 VNoRequest A Quote--
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller TypeMicroprocessor
DescriptionMultifuge X4R Pro 220V 60Hz
Capacity4 x 6 Microplates
Max. RCF7,197 x g Swing-Out with H-Flex HS4 rotor
Product LineMultifuge
RefrigeratedYes
DisplayFull Color Touch Screen Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesEnergy efficient, less waste
IncludesPrinted English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
MaterialFront, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Noise Level<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor18 Rotor options available
Rotor IncludedNo
Run TimeUp to 99 hr. 59 min. 59 sec.
StandardsISO 14971, ISO 13485, ISO 9001
Temperature Range-10°C to +40°C, ±2°C
TimerOption to count up or down
TypeBenchtop Centrifuge
VentilatedNo
Warranty5 Year Limited
Dimensions (L x W x H) Exterior690 x 746 x 361 mm
Length (English) Exterior27.2 in.
Length (Metric) Exterior690 mm
Width (English) Exterior29.4 in.
Width (Metric) Exterior746 mm
Height (English) Exterior14.2 in.
Height (English) Exterior Lid Open33.9 in.
Height (Metric) Exterior361 mm
Height (Metric) Exterior Lid Open861 mm
Depth (English)27.2 in.
Depth (Metric)690 mm
Net Weight120 kg
Voltage220 V
Wattage1950 w
Frequency60 Hz
Laboratory/Cleanroom UseCleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
Unit SizeEach
75009900Full specifications
EachMultifuge X4 Pro 208-240V 50/60Hz208/240 VNoRequest A Quote--
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller TypeMicroprocessor
DescriptionMultifuge X4 Pro 208-240V 50/60Hz
Capacity4 x 6 Microplates
Max. RCF7,197 x g Swing-Out with H-Flex HS4 rotor
Product LineMultifuge
RefrigeratedNo
DisplayFull Color Touch Screen Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesEnergy efficient, less waste
IncludesPrinted English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
MaterialFront, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Noise Level<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor18 Rotor options available
Rotor IncludedNo
Run TimeUp to 99 hr. 59 min. 59 sec.
StandardsISO 14971, ISO 13485, ISO 9001
TimerOption to count up or down
TypeBenchtop Centrifuge
VentilatedYes
Warranty5 Year Limited
Dimensions (L x W x H) Exterior690 x 566 x 362 mm
Length (English) Exterior27.2 in.
Length (Metric) Exterior690 mm
Width (English) Exterior22.3 in.
Width (Metric) Exterior566 mm
Height (English) Exterior14.2 in.
Height (English) Exterior Lid Open33.9 in.
Height (Metric) Exterior362 mm
Height (Metric) Exterior Lid Open861 mm
Depth (English)27.2 in.
Depth (Metric)690 mm
Net Weight89 kg
Voltage208/240 V
Wattage1600 w
Frequency50/60 Hz
Laboratory/Cleanroom UseCleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
Unit SizeEach
75009915Full specifications
EachMultifuge X4R Pro 220-240V 50Hz/230V 60Hz220/240 VNoRequest A Quote--
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller TypeMicroprocessor
DescriptionMultifuge X4R Pro 220-240V 50Hz/230V 60Hz
Capacity4 x 6 Microplates
Max. RCF7,197 x g Swing-Out with H-Flex HS4 rotor
Product LineMultifuge
RefrigeratedYes
DisplayFull Color Touch Screen Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesEnergy efficient, less waste
IncludesPrinted English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
MaterialFront, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Noise Level<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor18 Rotor options available
Rotor IncludedNo
Run TimeUp to 99 hr. 59 min. 59 sec.
StandardsISO 14971, ISO 13485, ISO 9001
Temperature Range-10°C to +40°C, ±2°C
TimerOption to count up or down
TypeBenchtop Centrifuge
VentilatedNo
Warranty5 Year Limited
Dimensions (L x W x H) Exterior690 x 746 x 361 mm
Length (English) Exterior27.2 in.
Length (Metric) Exterior690 mm
Width (English) Exterior29.4 in.
Width (Metric) Exterior746 mm
Height (English) Exterior14.2 in.
Height (English) Exterior Lid Open33.9 in.
Height (Metric) Exterior361 mm
Height (Metric) Exterior Lid Open861 mm
Depth (English)27.2 in.
Depth (Metric)690 mm
Net Weight120 kg
Voltage220/240 V
Wattage1950 w
Frequency50/60 Hz
Laboratory/Cleanroom UseCleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
Unit SizeEach
75016033Full specifications
EachMultifuge X4R Pro-MD, 120V TX-1000 Blood Tube Package120 VYes
Price: 20,000.00
Your price:
-
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2FCC part 15
Controller TypeMicroprocessor
DescriptionMultifuge X4R Pro-MD, 120V TX-1000 Blood Tube Package
Capacity196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes
Max. RCF4,122 x g (with TX-1000 rotor)
Max. Speed4,200 rpm (with TX-1000 rotor)
Product LineMultifuge
RefrigeratedYes
DisplayFull Color Touch Screen Interface
Drive SystemDirect, Brushless Induction Low Profile
Green FeaturesLess waste, Energy efficient
IncludesPrinted English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
MaterialFront, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
RotorBlood Tube Package Includes TX-1000 with Buckets, ClickSeal Biocontainment lids, 5/7 and 9/10 mL Blood Tube Adapters
Rotor IncludedYes
Run TimeUp to 99 hr. 59 min. 59 sec.
StandardsISO 14971, ISO 13485, ISO 9001
Temperature Range-10°C to +40°C, ±2°C
TimerOption to count up or down
TypeBenchtop Centrifuge
VentilatedNo
WarrantyCentrifuge: 5 Year Limited, Rotor: See Manual
Net WeightSee individual components of package
Voltage120 V
Frequency60 Hz
Laboratory/Cleanroom UseCleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
Unit SizeEach
Showing 10 of 18Show more
The GP Pro centrifuge series is available as ventilated models (X4) or refrigerated models (X4R) as well as for general laboratory use or IVD certified.
Performance features:
  • Intuitive touch screen user interface simplifies operation with easily accessible menu bar and centrifuge health icon for at-a-glance monitoring
  • Thermo Scientific™ Accumulated Centrifugal Effect (ACE™) integrator function for increased reproducibility
  • Thermo Scientific™ SMARTSpin™ imbalance technology optimizes safety and maximizes acceleration, braking and residual load imbalance for each rotor and bucket option
Thermo Scientific™ Auto-Lock™ Rotor Exchange:
  • Secure, ergonomic push-button rotor exchange in as little as three seconds with no tools required
  • Full chamber access for cleaning convenience and minimizes risk of contamination
  • Flexibility to quickly switch between applications and evolve changing laboratory needs
Thermo Scientific™ ClickSeal™ Biocontainment Lids:
  • Biocontainment sealing options for glove-friendly, one-handed open/close capability are ergonomically designed for both right and left-handed operations 
  • Audible and visual closure cues enhance safety and security
  • Biocontainment certification by National Institute for Health Protection, Porton Down, UK or TÜV NORD CERT GmbH, Germany
Rotor versatility and Thermo Scientific™ Fiberlite™ Carbon Fiber Rotors:
  • Up to 18 unique rotor options, including Fiberlite Carbon Fiber Rotor options, to help maximize performance and flexibility
  • Fiberlite carbon fiber rotors are supported by a 15-year warranty and no limitation in cycles of use
  • Carbon fiber offers a unique ability to mold each cavity to perfectly fit disposable conical tubes, bottles, and other consumables
Safety and ergonomic features:
  • Space saving design with compact dimensions to optimize work area at an ergonomic height 
  • Lid safety features promote ergonomic sample loading and unloading and enable easy cleaning
Energy efficiency:
  • System offers up to 40% energy consumption savings on industry standard protocols, such as standard blood separation or conical tube processing
  • Scheduling and compressor on/off capability promotes energy savings 
  • Fiberlite rotors weigh up to 60% less than metal equivalent rotors and therefore accelerate/decelerate faster translating into reduced energy consumption per run*  
Connectivity and data management:
  • Event Log, Rotor Log and Chart Data can be exported via USB in CSV or PDF formats
  • Programs are downloadable and shareable between units
  • Front USB port for easier plug in and data transfer
  • Optional Thermo Scientific™ SECURE-Spin™ Mode available, enables advanced and secure control features in the X Pro centrifuges by supporting 21 CFR Part 11 data management compliance and traceability 
    • Traceable documentation provides audit trail for setting and program changes
    • Compatible with Thermo Scientific™ Centri-Log™ Plus Data Management Software enabling remote monitoring of unlimited number of centrifuges and sample tracking with a barcode reader
Previous models
Multifuge X3, Multifuge X3R
*Compare data highlighted in the Green Fact Sheet from 2021 under https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/PG2030-PJT4622-COL33722-Centrifuges-FactSheet-Americas-FHR.pdf