Manostat™ Silicone Pumping Links
Actual product may vary
Thermo Scientific™
Manostat™ Silicone Pumping Links
Manostat silicone pumping links are for use with Manostat pumps and dispensers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTubing Size
7530016516
7230025525
7230017517
7530013513
7230013513
7230016516
7530014514
7530025525
7230014514
7530017517
Catalog number 75300165
Price (EUR)
-
Tubing Size:
16
Thermo Scientific™ Manostat silicone pumping links are for use with Manostat pumps and dispensers.
Specifications
For Use WithPreston, Spencer and Compulab RD1
Product LineThermo Scientific
Tubing Size16
Diameter (English) Inner1/8 in.
Diameter (English) Outer0.624 in.
Diameter (Metric) Inner3.2 mm
Diameter (Metric) Outer16 mm
MaterialSilicone
Product TypePumping Link
Unit SizeEach