MaxArray™ Breast Receptor Control Cell Block

Catalog number: 753023

 Related applications: Cell Structure | Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Full Product Description
MaxArray Breast Receptor Control Cell Block⁄Slides are formalin-fixed and paraffin-embedded using standard fixation methods. The MaxArray Breast Receptor Control Cell Block⁄Slides contains 4 cores: a negative control cell line, ER positive cell line, PR positive cell line and a cell line with HER2 overexpression. The cell lines will positively react to IHC staining of estrogen receptor, progesterone receptor and HER2 antibodies. The small surface area of the control block is advantageous as it allows side-by-side analysis of the tissue specimen and the control cell lines. For your convenience, the MaxArray Breast Receptor Control Cell Slides provide on slide control array staining which documents the staining pattern of your tissue sample for future reference. The MaxArray Breast Receptor Control Cell Block is ready to slice and use on regular microscope slides.

Applications: Immunohistochemistry, ISH, FISH, CISH
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Array Type: Tissue Microarrays
Format: Paraffin Cell Block
Sample Source: Cell Line
Tissue: Breast
Product Line: MaxArray™
Product Size: 1 block
Species: Human
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature.

Documents

Manuals & protocols