Manostat™ PharMed™ Pumping Links
Actual product may vary
Thermo Scientific™
Manostat™ PharMed™ Pumping Links
Manostat Pharmed pumping links are for use with Manostat pumps and dispensers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTubing SizeDescription
7530325525Preston, Spencer and Compulab RD1
7230313513Simon/Vera, Kate and Compulab RD2
7530313513Preston, Spencer and Compulab RD1
7230317517Simon/Vera, Kate and Compulab RD2
7530316516Preston, Spencer and Compulab RD1
7230325525Simon/Vera, Kate and Compulab RD2
7230316516Simon/Vera, Kate and Compulab RD2
7230314514Simon/Vera, Kate and Compulab RD2
7530317517Preston, Spencer and Compulab RD1
7530314514Preston, Spencer and Compulab RD1
Catalog number 75303255
Price (MXN)
-
Tubing Size:
25
Description:
Preston, Spencer and Compulab RD1
Thermo Scientific™ Manostat Pharmed pumping links are for use with Manostat pumps and dispensers.
Specifications
DescriptionPreston, Spencer and Compulab RD1
For Use WithPreston, Spencer, and Compulab RD1
Includes5 links/pack
Product LineThermo Scientific
Tubing Size25
Diameter (English) Inner3/16 in.
Diameter (English) Outer0.975 in.
Diameter (Metric) Inner4.8 mm
Diameter (Metric) Outer25 mm
MaterialPharMed BPT
Product TypePumping Link
Unit SizeEach