Manostat™ Tygon™ Pumping Links
Imagem ilustrativa. O produto pode variar
Thermo Scientific™
Manostat™ Tygon™ Pumping Links
Manostat Tygon pumping links are for use with Manostat pumps and dispensers.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoTubing SizeDescription
7531025525Preston, Spencer and Compulab RD1
7231017517Simon/Vera, Kate, Compulab RD2
7531014514Preston, Spencer and Compulab RD1
7531016516Preston, Spencer and Compulab RD1
7531017517Preston, Spencer and Compulab RD1
7231013513Simon/Vera, Kate, Compulab RD2
7231025525Simon/Vera, Kate, Compulab RD2
7231016516Simon/Vera, Kate, Compulab RD2
7231014514Simon/Vera, Kate, Compulab RD2
7531013513Preston, Spencer and Compulab RD1
Número do catálogo 75310255
Preço (BRL)
-
Tubing Size:
25
Description:
Preston, Spencer and Compulab RD1
Thermo Scientific™ Manostat Tygon pumping links are for use with Manostat pumps and dispensers.
Especificações
DescriptionPreston, Spencer and Compulab RD1
For Use WithPreston, Spencer and Compulab RD1
Includes5 links/pack
Product LineThermo Scientific
Tubing Size25
Diameter (English) Inner3/16 in.
Diameter (English) Outer0.975 in.
Diameter (Metric) Inner4.8 mm
Diameter (Metric) Outer25 mm
MaterialTygon
Product TypePump Tubing Links
Unit SizeEach